• 主页
  • 查找结果
  • Characterization and evaluation of 5-fluorouracil-loaded solid lipid nanoparticles prepared via a temperature-modulated solidification technique.
Characterization and evaluation of 5-fluorouracil-loaded solid lipid nanoparticles prepared via a temperature-modulated solidification technique.
AAPS PharmSciTech (2014-07-19)
Meghavi N Patel, Sushant Lakkadwala, Mohamed S Majrad, Elisha R Injeti, Steven M Gollmer, Zahoor A Shah, Sai Hanuman Sagar Boddu, Jerry Nesamony
摘要
The aim of this research was to advance solid lipid nanoparticle (SLN) preparation methodology by preparing glyceryl monostearate (GMS) nanoparticles using a temperature-modulated solidification process. The technique was reproducible and prepared nanoparticles without the need of organic solvents. An anticancer agent, 5-fluorouracil (5-FU), was incorporated in the SLNs. The SLNs were characterized by particle size analysis, zeta potential analysis, differential scanning calorimetry (DSC), infrared spectroscopy, atomic force microscopy (AFM), transmission electron microscopy (TEM), drug encapsulation efficiency, in vitro drug release, and in vitro cell viability studies. Particle size of the SLN dispersion was below 100 nm, and that of redispersed lyophilizates was ~500 nm. DSC and infrared spectroscopy suggested that the degree of crystallinity did not decrease appreciably when compared to GMS. TEM and AFM images showed well-defined spherical to oval particles. The drug encapsulation efficiency was found to be approximately 46%. In vitro drug release studies showed that 80% of the encapsulated drug was released within 1 h. In vitro cell cultures were biocompatible with blank SLNs but demonstrated concentration-dependent changes in cell viability to 5-FU-loaded SLNs. The 5-FU-loaded SLNs can potentially be utilized in an anticancer drug delivery system.
材料
货号
品牌
产品描述
Sigma-Aldrich
5-氟脲嘧啶, ≥99% (HPLC), powder
Sigma-Aldrich
N,N′-二琥珀酰亚胺碳酸酯, ≥95%
Sigma-Aldrich
聚四氟乙烯, powder (free-flowing), 1 μm particle size
Sigma-Aldrich
聚四氟乙烯, powder, >40 μm particle size
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
聚四氟乙烯, powder, 200 μm particle size
Sigma-Aldrich
N,N′-二琥珀酰亚胺碳酸酯, purum, ≥95.0% (NMR)
Sigma-Aldrich
聚四氟乙烯, beads
Supelco
5-氟脲嘧啶, analytical standard
Supelco
氟尿嘧啶, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
聚四氟乙烯, powder, 35 μm particle size
Sigma-Aldrich
聚四氟乙烯, powder (free-flowing), ≤12 μm particle size
USP
氟脲嘧啶, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
锗, chips, 99.999% trace metals basis
Sigma-Aldrich
聚四氟乙烯, powder, ≥350 μm particle size
氟尿嘧啶, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
氟脲嘧啶, meets USP testing specifications
锗, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
锗, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
锗, disks, 20mm, thickness 1.0mm, single crystal, 100%
锗, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
锗, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
锗, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
锗, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
锗, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
锗, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
锗, rod, 6mm, diameter 6.0mm, single crystal, 100%
锗, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
锗, sheet, 10x10mm, thickness 0.5mm, single crystal, -111, 100%