  • Search Results
  • Intracellular distribution and biological effects of phytochemicals in a sex steroid- sensitive model of human prostate adenocarcinoma.
Intracellular distribution and biological effects of phytochemicals in a sex steroid- sensitive model of human prostate adenocarcinoma.
Anti-cancer agents in medicinal chemistry (2014-06-26)
Antonella Smeriglio, Domenico Trombetta, Daniele Marcoccia, Laura Narciso, Alberto Mantovani, Stefano Lorenzetti
ABSTRACT
Prostate function is critical for male fertility and its well-known oncological biomarker, namely Prostate-Specific Antigen (PSA), can be also used to monitor prostate epithelial human cells upon treatment with pharmaceutical drugs or natural bioactive compounds. The LNCaP human prostate cell line was previously set up as a model system to investigate chemicals affecting prostate epithelium functionality by means of a tiered approach integrating two different toxicological endpoints, cell viability (MTS) and PSA secretion assays. Here, the same approach has been used to characterize the biological effects of phytochemicals on prostate epithelium. The antiandrogenic ability of phytochemicals to inhibit DHT-induced PSA secretion has been investigated also characterizing their intracellular distribution, in the presence or absence of sex steroids. Intracellular distribution allows to verify whether and to which extent each phytochemical is able to enter the cell and to reach the nucleus, the latter being the target of the supposed transcriptional modulatory activity upon phytochemicals' binding to sex steroid receptors. Some phytochemicals, supposed to have a role in the functionality of the prostate epithelium, have been tested in a dose-dependent manner in both MTS and PSA secretion assays. In parallel, to establish the "effective concentration", in comparison to the "nominal one", the intracellular amount of each phytochemical has been assessed upon cell fractionation of LNCaP-treated cells and subsequent chromatographic measurements.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
乙二胺四乙酸 二钠盐 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
槲皮素, ≥95% (HPLC), solid
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
白藜芦醇, ≥99% (HPLC)
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
4-(2-氨乙基)苯磺酰氟 盐酸盐, ≥97.0% (HPLC)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
染料木黄酮, synthetic, ≥98% (HPLC), powder
Supelco
Pefabloc® SC, analytical standard
Supelco
槲皮素, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
芹菜素, ≥95.0% (HPLC)
Sigma-Aldrich
(±)-柚皮素, ≥95%
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
乙二胺四乙酸, 99.995% trace metals basis
Supelco
白藜芦醇, certified reference material, TraceCERT®
USP
槲皮素, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
乙二胺四乙酸, BioUltra, ≥99.0% (KT)