  • Search Results
  • MicroRNA expression profiles associated with acquired gefitinib-resistance in human lung adenocarcinoma cells.
MicroRNA expression profiles associated with acquired gefitinib-resistance in human lung adenocarcinoma cells.
Molecular medicine reports (2014-10-24)
Xiaojun Ge, Limei Zheng, Min Huang, Yonglun Wang, Feng Bi
ABSTRACT
The aim of the present study was to establish, characterize and elucidate the potential mechanisms of acquired gefitinb resistance, using the A549 human lung cancer cell line. A gefitinib-resistant A549 sub-clone was established by exposure to escalating gefitinib concentrations over a period of 16-24 months. Half maximal inhibitory concentration (IC50) values were quantified using a real time cytotoxicity assay. The expression profiles of the parent and resistant sub-clone A549 cells were detected using the µParaflo® Microfluidics Biochip microRNA (miRNA) Microarray. The ArrayPro software was used to analyze the differential expression levels of the miRNA, and bioinformatics software was used to predict the potential target genes of the differentially expressed miRNAs. Quantitative polymerase chain reaction (qPCR) was used to confirm the results of the miRNA microarray. A miRNA mimic was transfected into the gefitinib-resistant cells, in order to predict target gene interaction effects, following gefitinib treatment. Protein expression level differences were confirmed by western-blot analysis. Real time cytotoxicity assays revealed a 3-fold increase in the IC50 values of the gefitinib-resistant sub-clones, as compared with the parent cells. There were marked morphological differences between the parent and resistant cells. In the microarray analysis, the gefitinib-resistant sub-clones had 25 upregulated and 18 downregulated miRNAs, as compared with the parent cells. The qPCR revealed that miR-7 was significantly downregulated, which was concordant with the results of the microarray. The results of the present study suggest that miR-7 may significantly improve the sensitivity of cancer cells to gefitinib. The data presented in the present study provides an experimental basis and theory that miRNAs may be involved in acquired gefitinib-resistance of lung adenocarcinoma, and miR-7 may have potential clinical effects in the reversal of drug resistance.
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
甘油, ≥99.5%
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
甘油, ACS reagent, ≥99.5%
Sigma-Aldrich
甘油, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸 二钠盐 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
苯甲磺酰氟, ≥98.5% (GC)
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
乙二醇-双(2-氨基乙醚)-N,N,N′,N′-四乙酸, for molecular biology, ≥97.0%
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
甘油, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Supelco
甘油, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
乙二醇-双(2-氨基乙醚)-N,N,N′,N′-四乙酸, ≥97.0%
Sigma-Aldrich
甘油, meets USP testing specifications
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
甘油 溶液, 83.5-89.5% (T)
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
甘油, BioXtra, ≥99% (GC)
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
甘油, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Supelco
氯化钠, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
甘油, FCC, FG