Merck
  • 主页
  • 查找结果
  • Sex-specific dose-response analysis of genotoxicity in cyproterone acetate-treated F344 rats.
Sex-specific dose-response analysis of genotoxicity in cyproterone acetate-treated F344 rats.
Mutation research. Genetic toxicology and environmental mutagenesis (2014-12-03)
Wei Ding, Michelle E Bishop, Mason G Pearce, Kelly J Davis, Gene A White, Lascelles E Lyn-Cook, Mugimane G Manjanatha
摘要
Cyproterone acetate (CPA), a synthetic hormonal drug, induces rat liver tumors in a sex-specific manner, with five-fold higher doses needed to induce liver tumors in male rats compared to females. In order to evaluate the potential of the in vivo alkaline Comet assay to predict the sex-specific carcinogenicity of CPA, CPA-induced direct DNA damage (DNA strand breaks and alkali-labile sites) were evaluated in the livers of both male and female F344 rats. In addition, secondary oxidative DNA damage was measured concurrently utilizing the human 8-oxoguanine-DNA-N-glycosylase (hOGG1) and EndonucleaseIII (EndoIII)-modified in vivo alkaline Comet assays and the reticulocyte micronucleus (MN) frequency was analyzed in peripheral blood. Groups of 5 seven-week-old male and female F344 rats received olive oil or 10, 25, 50 or 100 mg/kg bw CPA in olive oil by gavage at 0, 24, and 45 h and were sacrificed at 48 h. CPA-induced direct DNA damage in rat liver showed the same sex-specific pattern as its hepatotumorigenicity: a five-fold-higher dose of CPA was needed to induce a statistically significant increase in direct DNA damage in livers of males compared to females. However, peripheral blood MN frequency was weak in both sexes and CPA-induced oxidative DNA damage was generally greater in male than female rat livers. Taken together, our results demonstrate concordance in the sex-specificity of CPA in the in vivo alkaline Comet assay and cancer bioassay, while the induction of oxidative DNA damage by CPA was not directly correlated with its tumorigenicity.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
乙二胺四乙酸 二钠盐 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
甲醛 溶液, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
甲基磺酸甲酯, 99%
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
甲醛 溶液, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
乙二胺四乙酸, 99.995% trace metals basis
Supelco
甲醛 溶液, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
甲醛 溶液, tested according to Ph. Eur.
Sigma-Aldrich
乙二胺四乙酸, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
乙二胺四乙酸, ≥98.0% (KT)
Supelco
二甲基亚砜, analytical standard