• 主页
  • 查找结果
  • Effects of dietary macronutrient composition on exogenous neuropeptide Y's stimulation of food intake in chicks.
Effects of dietary macronutrient composition on exogenous neuropeptide Y's stimulation of food intake in chicks.
Neuroscience letters (2015-02-15)
Laura A Nelson, Elizabeth R Gilbert, Mark A Cline
摘要
In mammalian models it is well documented that the potent orexigenic factor, neuropeptide Y (NPY) causes preferential intake of high carbohydrate and fat diets; however, information on this is limited in non-mammalian species. The purpose of this study was to determine the influence of dietary macronutrient composition on NPY's orexigenic effect in chicks. Three isocaloric diets were formulated: high carbohydrate, fat and protein. In Experiment 1, chicks were fed the three diets and received intracerebroventricular injections of 0.2 or 2.0nmol NPY. Chicks that consumed the high carbohydrate and protein diets had a non-dose dependent similar magnitude of increased food intake after NPY injection, but those on the high fat diet had a dose dependent food intake increase. In Experiment 2, when chicks were given free access to all three diets, injection of 0.2nmol NPY caused preferential increase in intake of only the high protein diet whereas 2.0nmol NPY caused preferential increases in of both high carbohydrate and protein diets. Neither dose affected high fat diet intake. In Experiment 3, chicks were raised on one of the three diets and then switched to the others. When chicks were raised on the high fat and protein diets and then switched to the other diets, stimulation of food intake occurred for the same duration, 180min. However, when chicks were raised on the high carbohydrate and then switched to high fat, NPY injection caused a sustaining increase in cumulative food intake that lasted the entire observation period. These results suggest that NPY has selective effects on consumption of carbohydrate, fat and protein in chicks, and that diet in turn affects the NPY-mediated response in food intake, with a high fat diet enhancing NPY sensitivity that is associated with a greater magnitude and duration of feeding response. In turn, NPY caused preferential protein and carbohydrate intake instead of fat intake (in this order of preference), when chicks had the opportunity to select their diet. Finally, these results reinforce that NPY increases food intake independent of diet.
材料
货号
品牌
产品描述
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Supelco
胆钙化醇, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
胆钙化醇, ≥98% (HPLC)
Sigma-Aldrich
硒, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
硒, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Supelco
维生素D3标准液 CRM 溶液, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
维生素A标准液 溶液, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®
Supelco
胆钙化固醇(维生素 D3), analytical standard
胆钙化醇, European Pharmacopoeia (EP) Reference Standard
胆钙化醇, European Pharmacopoeia (EP) Reference Standard
USP
胆钙化醇, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
胆钙化醇, analytical standard
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
胆钙化醇, meets USP testing specifications
Sigma-Aldrich
硒, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
硒, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
硒, foil, 25x25mm, thickness 3mm, 99.95%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%