  • Search Results
  • Serum PCB levels in a representative sample of the Spanish adult population: the BIOAMBIENT.ES project.
Serum PCB levels in a representative sample of the Spanish adult population: the BIOAMBIENT.ES project.
The Science of the total environment (2014-07-08)
O Huetos, M Bartolomé, N Aragonés, M Cervantes-Amat, M Esteban, M Ruiz-Moraga, B Pérez-Gómez, E Calvo, M Vila, A Castaño
ABSTRACT
This manuscript presents the levels of six indicator polychlorinated biphenyl (PCB) congeners (IUPAC nos. 28, 52, 101, 138, 153 and 180) in the serum of 1880 individuals from a representative sample of the Spanish working population recruited between March 2009 and July 2010. Three out of the six PCBs studied (180, 153 and 138) were quantified in more than 99% of participants. PCB 180 was the highest contributor, followed by PCBs 153 and 138, with relative abundances of 42.6%, 33.2% and 24.2%, respectively. In contrast, PCBs 28 and 52 were detected in only 1% of samples, whereas PCB 101 was detectable in 6% of samples. The geometric mean (GM) for ΣPCBs138/153/180 was 135.4 ng/g lipid (95% CI: 121.3-151.2 ng/g lipid) and the 95th percentile was 482.2 ng/g lipid. Men had higher PCB blood concentrations than women (GMs 138.9 and 129.9 ng/g lipid respectively). As expected, serum PCB levels increased with age and frequency of fish consumption, particularly in those participants younger than 30 years of age. The highest levels we found were for participants from the Basque Country, whereas the lowest concentrations were found for those from the Canary Islands. The Spanish population studied herein had similar levels to those found previously in Greece and southern Italy, lower levels than those in France and central Europe, and higher PCB levels than those in the USA, Canada and New Zealand. This paper provides the first baseline information regarding PCB exposure in the Spanish adult population on a national scale. The results will allow us to establish reference levels, follow temporal trends and identify high-exposure groups, as well as monitor implementation of the Stockholm Convention in Spain.
Sigma-Aldrich
硫酸, ACS reagent, 95.0-98.0%
Sigma-Aldrich
甲酸, reagent grade, ≥95%
Sigma-Aldrich
硫酸, 99.999%
Sigma-Aldrich
甲酸, ACS reagent, ≥96%
Sigma-Aldrich
甲酸, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Supelco
硫酸 溶液, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
2,2,4-三甲基戊烷, anhydrous, 99.8%
Sigma-Aldrich
2,2,4-三甲基戊烷, ACS reagent, ≥99.0%
Sigma-Aldrich
2,2,4-三甲基戊烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Supelco
硫酸, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
硫酸, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
2,2,4-三甲基戊烷, suitable for HPLC, ≥99%
Sigma-Aldrich
甲酸, ACS reagent, ≥88%
Sigma-Aldrich
2,2,4-三甲基戊烷, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
甲酸, ≥95%, FCC, FG
Sigma-Aldrich
硫酸, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
甲酸 溶液, BioUltra, 1.0 M in H2O
Sigma-Aldrich
硫酸 溶液, puriss. p.a., ≥25% (T)
Supelco
2,2,4-三甲基戊烷, analytical standard
Sigma-Aldrich
2,2,4-三甲基戊烷, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,4-三甲基戊烷, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
密度标准品 692kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
硫酸, ACS reagent, 95.0-98.0%