• 主页
  • 查找结果
  • Manganese superoxide dismutase is required to maintain osteoclast differentiation and function under static force.
Manganese superoxide dismutase is required to maintain osteoclast differentiation and function under static force.
Scientific reports (2015-01-27)
Tao Guo, Liqiang Zhang, Anna Konermann, Hong Zhou, Fang Jin, Wenjia Liu
摘要
Bone homeostasis is maintained by the balance of osteoblasts (OBs) and osteoclasts (OCs). Increased activity of OCs not only contributes to pathological bone resorption, such as osteoporosis and periodontitis, but also is responsible for physiological conditions like orthodontic tooth movement (OTM). However, the detailed mechanism by which orthodontic force promotes the formation of OCs is still poorly understood. In this study, we confirmed that static force promoted the differentiation of human cord monocytes (HMNCs) into OCs depending on loading time and magnitude. Protein expression profiles among HMNCs, HMNCs subjected to static force and mature OCs were established via 2-DE and MALDI-TOF-MS analyses. Total respective protein spot numbers of 549 ± 13, 612 ± 19 and 634 ± 16 were detected in each of the gels by image analysis. The five proteins identified were plasminogen activator inhibitor 2 (PAI-2, Spot 1), peroxiredoxin-6 (PRD-6, Spot 3), manganese superoxide dismutase (SOD2, Spot 6), Rho GDP-dissociation inhibitor 2 (Rho-GDI2, Spot 11) and L-lactate dehydrogenase B chain (L-LDH, Spot 15). More importantly, we revealed that SOD2 was required to maintain monocyte differentiation into functional OCs and may become a potential target for regulating the efficiency of OTM in the future.
材料
货号
品牌
产品描述
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
丙酮, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
噻唑蓝, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
噻唑蓝, 98%
Sigma-Aldrich
丙酮, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-谷氨酰胺, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
丙酮, Laboratory Reagent, ≥99.5%
SAFC
L-谷氨酰胺
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.8%
Supelco
丙酮, analytical standard
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Supelco
丙酮, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
丙酮, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
二喹啉甲酸 二钠盐 水合物, ≥98% (HPLC)
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
丙酮, histological grade, ≥99.5%
Sigma-Aldrich
L-谷氨酰胺, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
丙酮, natural, ≥97%
Sigma-Aldrich
L-谷氨酰胺, γ-irradiated, BioXtra, suitable for cell culture
Supelco
丙酮 溶液, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
USP
丙酮, United States Pharmacopeia (USP) Reference Standard
Supelco
L-谷氨酰胺, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
丙酮, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
Supelco
L-谷氨酰胺, certified reference material, TraceCERT®
Sigma-Aldrich
L-谷氨酰胺