Merck
Effects of cerebrolysin on rat Schwann cells in vitro.
Acta histochemica (2014-03-19)
Benjamin Lucas, Josephine Pinkernelle, Hisham Fansa, Gerburg Keilhoff
ABSTRACT
Although the peripheral nervous system (PNS) is capable of regeneration, these processes are limited. As a potential means to augment PNS regeneration, the effects of cerebrolysin (CL), a proteolytic peptide fraction, were tested in vitro on Schwann cell (SC) proliferation, stress resistance, phagocytic and cluster-forming capacity. Primary SC/fibrocyte co-cultures were prepared from dorsal root ganglia of 5-7-day-old rats. SCs were subjected to mechanical stress by media change and metabolic stress by serum glucose deprivation (SGD). Cell survival was assessed using MTT test. SC proliferation was determined by counting BrdU-labeled cells. SC clustering was studied by ImageJ analysis of S100 immunostaining. Wallerian degeneration (WD) was evaluated by measuring acetylcholine-esterase staining within sciatic nerves in vitro. It was found that CL caused no effect on MTT turnover in the tested doses. CL inhibited SC proliferation in a dose-dependent manner. Media change and additional SGD stress inhibited SC clustering. CL enhanced the reorganization of SC clusters and was able to counteract SGD-induced cluster defects. Moreover, CL accelerated WD in vitro. CL was able to enhance the functions of SCs that are relevant to nerve regeneration. Thus, our findings suggest that CL may be suitable for therapeutic usage to enhance PNS regeneration/reconstruction.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
蔗糖, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
蔗糖, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
蔗糖, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
蔗糖, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
蔗糖, ACS reagent
USP
蔗糖, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, ≥99.5%
Sigma-Aldrich
蔗糖, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
蔗糖, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
蔗糖, Grade II, suitable for plant cell culture
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
蔗糖, Grade I, suitable for plant cell culture
Sigma-Aldrich
蔗糖, meets USP testing specifications
Supelco
二甲基亚砜, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
二甲基亚砜, analytical standard
USP
二甲基亚砜, United States Pharmacopeia (USP) Reference Standard
蔗糖, European Pharmacopoeia (EP) Reference Standard