  • Search Results
  • Delivery of berberine using chitosan/fucoidan-taurine conjugate nanoparticles for treatment of defective intestinal epithelial tight junction barrier.
Delivery of berberine using chitosan/fucoidan-taurine conjugate nanoparticles for treatment of defective intestinal epithelial tight junction barrier.
Marine drugs (2014-11-26)
Shao-Jung Wu, Trong-Ming Don, Cheng-Wei Lin, Fwu-Long Mi
ABSTRACT
Bacterial-derived lipopolysaccharides (LPS) can cause defective intestinal barrier function and play an important role in the development of inflammatory bowel disease. In this study, a nanocarrier based on chitosan and fucoidan was developed for oral delivery of berberine (Ber). A sulfonated fucoidan, fucoidan-taurine (FD-Tau) conjugate, was synthesized and characterized by Fourier transform infrared (FTIR) spectroscopy. The FD-Tau conjugate was self-assembled with berberine and chitosan (CS) to form Ber-loaded CS/FD-Tau complex nanoparticles with high drug loading efficiency. Berberine release from the nanoparticles had fast release in simulated intestinal fluid (SIF, pH 7.4), while the release was slow in simulated gastric fluid (SGF, pH 2.0). The effect of the berberine-loaded nanoparticles in protecting intestinal tight-junction barrier function against nitric oxide and inflammatory cytokines released from LPS-stimulated macrophage was evaluated by determining the transepithelial electrical resistance (TEER) and paracellular permeability of a model macromolecule fluorescein isothiocyanate-dextran (FITC-dextran) in a Caco-2 cells/RAW264.7 cells co-culture system. Inhibition of redistribution of tight junction ZO-1 protein by the nanoparticles was visualized using confocal laser scanning microscopy (CLSM). The results suggest that the nanoparticles may be useful for local delivery of berberine to ameliorate LPS-induced intestinal epithelia tight junction disruption, and that the released berberine can restore barrier function in inflammatory and injured intestinal epithelial.
Sigma-Aldrich
1,2-二氯乙烷, anhydrous, 99.8%
Sigma-Aldrich
噻唑蓝, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
噻唑蓝, 98%
Sigma-Aldrich
2',7'-二氯荧光素二乙酸酯, ≥97%
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
1,2-二氯乙烷, ACS reagent, ≥99.0%
Sigma-Aldrich
荧光素异硫氰酸酯异构体I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
荧光素 5(6)-异硫氰酸酯, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
1,2-二氯乙烷, suitable for HPLC, ≥99.8%
Sigma-Aldrich
牛磺酸, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
MES 溶液, BioUltra, for molecular biology, 0.5 M in H2O
Supelco
1,2-二氯乙烷, analytical standard
Supelco
牛磺酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
荧光素异硫氰酸酯异构体I, ≥97.5% (HPLC)
Sigma-Aldrich
荧光素 5(6)-异硫氰酸酯, ≥90% (HPLC)
Sigma-Aldrich
牛磺酸, BioUltra, ≥99.5% (T)
SAFC
牛磺酸
Sigma-Aldrich
牛磺酸, ≥98%, FG
USP
牛磺酸, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
荧光素异硫氰酸酯异构体I, ≥97.5% (HPLC)