Sigma-Aldrich
乙酸, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
乙酸, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
乙酸, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
地塞米松, ≥98% (HPLC), powder
Sigma-Aldrich
吲哚美辛, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
乙酸, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
甲醛 溶液, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
乙酸 溶液, suitable for HPLC
Sigma-Aldrich
乙酸, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Supelco
乙酸, analytical standard
Supelco
地塞米松, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
乙酸, natural, ≥99.5%, FG
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)
Sigma-Aldrich
甲醛 溶液, meets analytical specification of USP, ≥34.5 wt. %
USP
冰醋酸, United States Pharmacopeia (USP) Reference Standard
Supelco
甲醛 溶液, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
地塞米松, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
乙酸, ≥99.5%, FCC, FG
Supelco
吲哚美辛, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
甲醛 溶液, tested according to Ph. Eur.
Sigma-Aldrich
乙酸, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
地塞米松, meets USP testing specifications
Sigma-Aldrich
吲哚美辛, meets USP testing specifications
地塞米松, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
乙酸, glacial, puriss., 99-100%