• 主页
  • 查找结果
  • Correlation between sperm parameters and protein expression of antioxidative defense enzymes in seminal plasma: a pilot study.
Correlation between sperm parameters and protein expression of antioxidative defense enzymes in seminal plasma: a pilot study.
Disease markers (2015-02-19)
Biljana Macanovic, Milica Vucetic, Aleksandra Jankovic, Ana Stancic, Biljana Buzadzic, Eliana Garalejic, Aleksandra Korac, Bato Korac, Vesna Otasevic
摘要
Semen analysis is the cornerstone in the evaluation of male (in)fertility. However, there are men with normal semen tests but with impaired fertilizing ability, as well as fertile men with poor sperm characteristics. Thus, there is rising interest to find novel parameters that will help to predict and define the functional capacity of spermatozoa. We examined whether there is a correlation between semen parameters (count, progressive motility, and morphology) and protein expression/activity of antioxidative defense enzymes in seminal plasma from 10 normospermic subjects. Sperm progressive motility was in positive correlation with seminal plasma protein expression of both superoxide dismutase (SOD) isoforms (MnSOD and CuZnSOD) and catalase. Also, positive correlation was observed between sperm count and MnSOD protein expression, as well as between sperm morphology and protein expression of catalase in seminal plasma. In contrast, protein expression of glutathione peroxidase was not in correlation with any sperm parameter, while its activity negatively correlated with sperm morphology and motility. These data suggest that evaluation of protein expression of antioxidative defense enzymes in seminal plasma might be of importance in the evaluation of male fertility status and that could be used as an additional biomarker along with classic semen analysis in assessment of semen quality.
材料
货号
品牌
产品描述
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%