  • Search Results
  • Phytol, a diterpene alcohol, inhibits the inflammatory response by reducing cytokine production and oxidative stress.
Phytol, a diterpene alcohol, inhibits the inflammatory response by reducing cytokine production and oxidative stress.
Fundamental & clinical pharmacology (2013-10-10)
Renan O Silva, Francisca Beatriz M Sousa, Samara R B Damasceno, Nathalia S Carvalho, Valdelânia G Silva, Francisco Rodrigo M A Oliveira, Damião P Sousa, Karoline S Aragão, André L R Barbosa, Rivelilson M Freitas, Jand Venes R Medeiros
ABSTRACT
Studies have shown that diterpenes have anti-inflammatory and redox-protective pharmacological activities. The present study aimed to investigate the anti-inflammatory properties of phytol, a diterpene alcohol, in a mouse model of acute inflammation, and phytol effect on leukocyte recruitment, cytokines levels, and oxidative stress. The anti-inflammatory activities of phytol were assessed by measuring paw edema induced by different inflammatory agents (e.g., λ-carrageenan, compound 48/80, histamine, serotonin, bradykinin, and prostaglandin E2 [PGE2 ]), myeloperoxidase (MPO) activity, peritonitis model and cytokine levels. Further, oxidative stress was evaluated by determining glutathione (GSH) levels and malondialdehyde (MDA) concentration. The results showed that phytol (7.5, 25, 50, and 75 mg/kg) significantly reduced carrageenan-induced paw edema, in a dose-dependent manner. In addition, phytol (75 mg/kg) inhibited compound 48/80-, histamine-, serotonin-, bradykinin- and PGE2 -induced paw edema. It also inhibited the recruitment of total leukocytes and neutrophils; decreased MPO activity, tumor necrosis factor-α (TNF-α) and interleukin-1β (IL-1β) levels, and MDA concentration; and increased GSH levels during carrageenan-induced acute inflammation. These results suggest that phytol attenuates the inflammatory response by inhibiting neutrophil migration that is partly caused by reduction in IL-1β and TNF-α levels and oxidative stress.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
吲哚美辛, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
组胺, ≥97.0%
Sigma-Aldrich
前列腺素E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
前列腺素E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
前列腺素E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
组胺, analytical standard
Supelco
吲哚美辛, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
叶绿醇, ≥97%, FG
Supelco
二甲基亚砜, analytical standard
Supelco
二甲基亚砜, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
二甲基亚砜, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
吲哚美辛, meets USP testing specifications
Sigma-Aldrich
叶绿醇, 97%, mixture of isomers
二甲基亚砜, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
二甲基亚砜, puriss. p.a., dried, ≤0.02% water
USP
吲哚美辛, United States Pharmacopeia (USP) Reference Standard
吲哚美辛, European Pharmacopoeia (EP) Reference Standard
前列腺素E2, European Pharmacopoeia (EP) Reference Standard