  • Search Results
  • Isolation and characterization of marine Brevibacillus sp. S-1 collected from South China Sea and a novel antitumor peptide produced by the strain.
Isolation and characterization of marine Brevibacillus sp. S-1 collected from South China Sea and a novel antitumor peptide produced by the strain.
PloS one (2014-11-06)
Lanhong Zheng, Yao Yi, Jia Liu, Xiukun Lin, Kangli Yang, Mei Lv, Xinwen Zhou, Jianhua Hao, Junzhong Liu, Yuan Zheng, Mi Sun
ABSTRACT
A Gram-positive, rod-shaped bacterium, designated as S-1, was isolated from a marine sediment sample collected from South China Sea. Phylogenetic analysis based on 16S rRNA gene sequence showed that S-1 belongs to the genus Brevibacillus. A novel cytotoxic peptide was isolated from the fermentation broth of the marine-derived bacterium Brevibacillus sp. S-1, using ion-exchange chromatography and reverse-phase HPLC chromatography. The molecular weight of this peptide was determined as 1570 Da by MALDI-TOF mass spectrometry, and its structure was proposed as a cyclic peptide elucidated by MALDI-TOF/TOF mass spectrometry and de novo sequencing. 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide (MTT) assay showed that this peptide exhibited cytotoxicity against BEL-7402 human hepatocellular carcinoma cells, RKO human colon carcinoma cells, A549 human lung carcinoma cells, U251 human glioma cells and MCF-7 human breast carcinoma cells. Additionally, SBP exhibited low cytotoxicity against HFL1 human normal fibroblast lung cells. The result suggested that the cytotoxic effect of the peptide is specific to tumor cells.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
焦碳酸二乙酯, 96% (NT)
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
焦碳酸二乙酯, 96% (NT)
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
焦碳酸二乙酯, 96% (NT)
Supelco
二甲基亚砜, analytical standard
Supelco
二甲基亚砜, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
二甲基亚砜, United States Pharmacopeia (USP) Reference Standard
二甲基亚砜, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
二甲基亚砜, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
二甲基亚砜, ≥99.6%, ReagentPlus®