Sigma-Aldrich
二甲基亚砜, Hybri-Max™, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
氯化镁 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
氯化镁, anhydrous, ≥98%
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
氯化镁, powder, <200 μm
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
氯化镁, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
氯化镁 溶液, PCR Reagent, 25 mM MgCI2 solution for PCR
Supelco
二甲基亚砜, analytical standard
Sigma-Aldrich
氯化镁, AnhydroBeads™, −10 mesh, 99.99% trace metals basis
Supelco
二甲基亚砜, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
二甲基亚砜, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
氯化镁, AnhydroBeads™, −10 mesh, 99.9% trace metals basis
二甲基亚砜, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
二甲基亚砜, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
氯化镁 溶液, 0.1 M
Sigma-Aldrich
二甲基亚砜, ≥99.6%, ReagentPlus®