  • Search Results
  • Regulation of lipid production by light-emitting diodes in human sebocytes.
Regulation of lipid production by light-emitting diodes in human sebocytes.
Archives of dermatological research (2015-02-19)
Yu Ra Jung, Sue Jeong Kim, Kyung Cheol Sohn, Young Lee, Young Joon Seo, Young Ho Lee, Kyu Uang Whang, Chang Deok Kim, Jeung Hoon Lee, Myung Im
ABSTRACT
Light-emitting diodes (LED) have been used to treat acne vulgaris. However, the efficacy of LED on sebaceous lipid production in vitro has not been examined. This study investigated the efficacy of 415 nm blue light and 630 nm red light on lipid production in human sebocytes. When applied to human primary sebocytes, 415 nm blue light suppressed cell proliferation. Based on a lipogenesis study using Oil Red O, Nile red staining, and thin-layered chromatography, 630 nm red light strongly downregulated lipid production in sebocytes. These results suggest that 415 nm blue light and 630 nm red light influence lipid production in human sebocytes and have beneficial effects on acne by suppressing sebum production.
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
丙酮, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
丙酮, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.8%
Sigma-Aldrich
丙酮, Laboratory Reagent, ≥99.5%
Supelco
丙酮, analytical standard
Supelco
丙酮, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
USP
丙酮, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
丙酮, histological grade, ≥99.5%
Sigma-Aldrich
丙酮, natural, ≥97%
Sigma-Aldrich
丙酮, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
丙酮 溶液, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
丙酮, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
丙酮, purum, ≥99.0% (GC)