• 主页
  • 查找结果
  • Ischemic postconditioning relieves cerebral ischemia and reperfusion injury through activating T-LAK cell-originated protein kinase/protein kinase B pathway in rats.
Ischemic postconditioning relieves cerebral ischemia and reperfusion injury through activating T-LAK cell-originated protein kinase/protein kinase B pathway in rats.
Stroke (2014-07-12)
Haiping Zhao, Rongliang Wang, Zhen Tao, Li Gao, Feng Yan, Zhi Gao, Xiangrong Liu, Xunming Ji, Yumin Luo
摘要
Ischemic postconditioning (IPostC) protects against ischemic brain injury. To date, no study has examined the role of T-LAK-cell-originated protein kinase (TOPK) in IPostC-afforded neuroprotection. We explored the molecular mechanism related with TOPK in antioxidant effect of IPostC against ischemia/reperfusion. Focal ischemia was induced in rats by transient middle cerebral artery occlusion. Reactive oxygen species production in the peri-infarct cortex was detected using dihydroethidium. Malondialdehyde, as a marker of lipid peroxidation, and 3-nitrotyrosine, as a marker of protein oxidation, were detected by ELISA. The expression or location of antioxidant proteins and signal molecules TOPK, phosphatase, and tensin homolog, and Akt was analyzed by Western blotting and immunofluorescence. Our results revealed that IPostC relieved transient middle cerebral artery occlusion-induced oxidative damage by reducing reactive oxygen species, malondialdehyde, and 3-nitrotyrosine accumulation in the peri-infarct cortex and raised levels of antioxidants perioxiredoxin-1, peroxiredoxin-2, and thioredoxin-1. In addition, IPostC increased p-AKT and p-TOPK levels, which colocalized in neural cells. In vitro TOPK knockdown by small interfering RNA decreased the levels of antioxidants peroxiredoxin-1, thioredoxin, and manganese superoxide dismutase activity in PC12 cells. In vivo intracerebroventricular injection of TOPK small interfering RNA reversed IPostC-induced neuroprotection by increasing infarct volume and nitric oxide content and reducing manganese superoxide dismutase activity. Moreover, IPostC-evoked Akt activation was blocked by TOPK small interfering RNA in vivo, but the decreased phosphorylated phosphatase and tensin homolog level in ischemia/reperfusion was not influenced by IPostC or by TOPK small interfering RNA treatment. Our results suggest that the antioxidative effects of TOPK/Akt might contribute to the neuroprotection of IPostC treatment against transient middle cerebral artery occlusion.
材料
货号
品牌
产品描述
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
3-硝基-L-酪氨酸, crystalline
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%