  • Search Results
  • Inhibitory effect of Ecliptae herba extract and its component wedelolactone on pre-osteoclastic proliferation and differentiation.
Inhibitory effect of Ecliptae herba extract and its component wedelolactone on pre-osteoclastic proliferation and differentiation.
Journal of ethnopharmacology (2014-10-01)
Yan-Qiu Liu, Li-Bin Zhan, Tiegang Liu, Meng-Chun Cheng, Xiao-Yan Liu, Hong-Bin Xiao
ABSTRACT
Ecliptae herba, also known as "Mo-Han-Lian", has long been used in China to nourish Kidney and thereafter strengthen bones. Accumulating evidence indicates that extracts of Ecliptae herba have antiosteoporotic effect. However, the effective compounds and cellular mode of action are still unclear. To investigate the effect of ethyl acetate extract of Ecliptae herba (EAE) and its component wedelolactone on proliferation and differentiation of preosteoclastic RAW264.7 cells as well as proliferation of bone marrow stromal cells (BMSC). RAW264.7 and BMSC were examined for proliferation by a 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2-H-tetrazolium bromide (MTT) method. Tartrate-resistant acid phosphatase (TRAP) activity of RAW264.7 was measured by using p-nitrophenyl sodium phosphate (pNPP) assay after the cells were treated with 30ng/ml receptor activator for nuclear factor-κ B ligand (RANKL) plus various concentrations of EAE, wedelolactone or alendronate. The formation of multinucleated TRAP-positive RAW264.7 cells was observed by using a TRAP-staining kit. Treatment of RAW264.7 cells with EAE at high doses (20µg/ml and 40µg/ml) or wedelolactone at 10µg/ml resulted in a decrease in proliferation of RAW264.7 cells. Low doses of EAE (5, 10µg/ml) and wedelolactone (2.5µg/ml) inhibited RANKL-induced TRAP activity by 20.3%, 37.9%, and 48.3%. The inhibitory effect of wedelolactone is more potent than that of alendronate, an anti-resorptive drug. Morphological changes revealed that 5µg/ml EAE and 2.5µg/ml wedelolactone reduced the number of multinucleated osteoclast-like cells. At the high doses, EAE (20µg/ml) and wedelolactone (10µg/ml) inhibited the growth of BMSC. EAE and its component wedelolactone inhibited osteoclast RAW264.7 proliferation and differentiation at the low doses, but at the high doses, showed cytotoxic effect on BMSC. These results indicated that EAE and wedelolatone might be potential alternative therapy for osteoporosis.
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
丙酮, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
乙腈, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
丙酮, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.8%
Sigma-Aldrich
丙酮, Laboratory Reagent, ≥99.5%
Supelco
丙酮, analytical standard
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
丙酮, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
乙腈, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Supelco
乙腈, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
USP
丙酮, United States Pharmacopeia (USP) Reference Standard
Supelco
乙腈, analytical standard
Sigma-Aldrich
丙酮, histological grade, ≥99.5%
Sigma-Aldrich
丙酮, natural, ≥97%
Sigma-Aldrich
丙酮, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
丙酮 溶液, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
丙酮, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
乙腈, ReagentPlus®, 99%