• 主页
  • 查找结果
  • Acute dietary tryptophan manipulation differentially alters social behavior, brain serotonin and plasma corticosterone in three inbred mouse strains.
Acute dietary tryptophan manipulation differentially alters social behavior, brain serotonin and plasma corticosterone in three inbred mouse strains.
Neuropharmacology (2014-12-03)
Wynne Q Zhang, Corey M Smolik, Priscilla A Barba-Escobedo, Monica Gamez, Jesus J Sanchez, Martin A Javors, Lynette C Daws, Georgianna G Gould
摘要
Clinical evidence indicates brain serotonin (5-HT) stores and neurotransmission may be inadequate in subpopulations of individuals with autism, and this may contribute to characteristically impaired social behaviors. Findings that depletion of the 5-HT precursor tryptophan (TRP) worsens autism symptoms support this hypothesis. Yet dietetic studies show and parents report that many children with autism consume less TRP than peers. To measure the impact of dietary TRP content on social behavior, we administered either diets devoid of TRP, with standard TRP (0.2 g%), or with 1% added TRP (1.2 g%) overnight to three mouse strains. Of these, BTBRT(+)Itpr3(tf)/J and 129S1/SvImJ consistently exhibit low preference for social interaction relative to C57BL/6. We found that TRP depletion reduced C57BL/6 and 129S social interaction preference, while TRP enhancement improved BTBR sociability (p < 0.05; N = 8-10). Subsequent marble burying did not differ among diets or strains. After behavior tests, brain TRP levels and plasma corticosterone were higher in TRP enhanced C57BL/6 and BTBR, while 5-HT levels were reduced in all strains by TRP depletion (p < 0.05; N = 4-10). Relative hyperactivity of BTBR and hypoactivity of 129S, evident in self-grooming and chamber entries during sociability tests, were uninfluenced by dietary TRP. Our findings demonstrate mouse sociability and brain 5-HT turnover are reduced by acute TRP depletion, and can be enhanced by TRP supplementation. This outcome warrants further basic and clinical studies employing biomarker combinations such as TRP metabolism and 5-HT regulated hormones to characterize conditions wherein TRP supplementation may best ameliorate sociability deficits.
材料
货号
品牌
产品描述
Sigma-Aldrich
碘, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
乙二胺四乙酸 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
乙二胺四乙酸, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
碘, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
肾上腺酮, ≥98.5% (HPLC)
Sigma-Aldrich
乙二胺四乙酸, 99.995% trace metals basis
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
肾上腺酮, ≥92%
Sigma-Aldrich
5-羟色胺肌酸酐硫酸盐 一水合物, powder
Sigma-Aldrich
乙二胺四乙酸, ≥98.0% (KT)
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
乙二胺四乙酸, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
Supelco
肾上腺酮, VETRANAL®, analytical standard
Sigma-Aldrich
血清素 马来酸氢盐, powder
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%