Merck
  • 主页
  • 查找结果
  • Evaluation of spin freezing versus conventional freezing as part of a continuous pharmaceutical freeze-drying concept for unit doses.
Evaluation of spin freezing versus conventional freezing as part of a continuous pharmaceutical freeze-drying concept for unit doses.
International journal of pharmaceutics (2015-05-20)
L De Meyer, P-J Van Bockstal, J Corver, C Vervaet, J P Remon, T De Beer
摘要
Spin-freezing as alternative freezing approach was evaluated as part of an innovative continuous pharmaceutical freeze-drying concept for unit doses. The aim of this paper was to compare the sublimation rate of spin-frozen vials versus traditionally frozen vials in a batch freeze-dryer, and its impact on total drying time. Five different formulations, each having a different dry cake resistance, were tested. After freezing, the traditionally frozen vials were placed on the shelves while the spin-frozen vials were placed in aluminum vial holders providing radial energy supply during drying. Different primary drying conditions and chamber pressures were evaluated. After 2h of primary drying, the amount of sublimed ice was determined in each vial. Each formulation was monitored in-line using NIR spectroscopy during drying to determine the sublimation endpoint and the influence of drying conditions upon total drying time. For all tested formulations and applied freeze-drying conditions, there was a significant higher sublimation rate in the spin-frozen vials. This can be explained by the larger product surface and the lower importance of product resistance because of the much thinner product layers in the spin frozen vials. The in-line NIR measurements allowed evaluating the influence of applied drying conditions on the drying trajectories.
材料
货号
品牌
产品描述
Sigma-Aldrich
甘氨酸, suitable for electrophoresis, ≥99%
Sigma-Aldrich
甘氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
高氯酸, ACS reagent, 70%
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-组氨酸, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
甘氨酸, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
高氯酸, 70%, 99.999% trace metals basis
Sigma-Aldrich
甘氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
L-组氨酸
Sigma-Aldrich
甘氨酸, ACS reagent, ≥98.5%
Sigma-Aldrich
高氯酸, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
SAFC
甘氨酸
Sigma-Aldrich
甘氨酸, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
高氯酸, ACS reagent, 60%
Sigma-Aldrich
甘氨酸, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-组氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
甘氨酸, BioXtra, ≥99% (titration)
Sigma-Aldrich
甘氨酸, 99%, FCC
Sigma-Aldrich
高氯酸, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)