  • Search Results
  • Novel role of hnRNP-A2/B1 in modulating aryl hydrocarbon receptor ligand sensitivity.
Novel role of hnRNP-A2/B1 in modulating aryl hydrocarbon receptor ligand sensitivity.
Archives of toxicology (2014-09-17)
See-Wun Cho, Ken-ichi Suzuki, Yoshiaki Miura, Tatsuhiko Miyazaki, Masato Nose, Hisato Iwata, Eun-Young Kim
ABSTRACT
The aryl hydrocarbon receptor (AHR) is responsible for susceptibility to its ligand-dependent responses. However, the effect of non-AHR factors is less clear. To explore the non-AHR factors, we used two mouse strains with different AHR genetic variants, namely C3H/lpr and MRL/lpr strains with Ala and Val as the 375th amino acid residue, respectively. To assess the contribution of AHR alone, COS-7 cells transiently expressing AHR from each strain were treated with 6-formylindolo[3,2-b]carbazole (FICZ) and 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD), and xenobiotic-responsive element (XRE)-driven reporter gene activities were measured. FICZ-EC50 values for the C3H/lpr and MRL/lpr AHR-mediated transactivation were 0.023 and 0.046 nM, respectively, indicating a similar susceptibility in both AHR genotypes. In contrast, C3H/lpr AHR was fourfold more sensitive to TCDD than MRL/lpr AHR. By a pull-down assay using a XRE-containing PCR product as bait and the hepatic nuclear extracts of both FICZ-treated mouse strains, we identified two interacting proteins as heterogeneous nuclear ribonucleoprotein A2/B1 (hnRNP-A2) and its splicing variant (hnRNP-A2b). Immunoprecipitation assays demonstrated the AHR interaction with hnRNP-A2/B1. When hnRNP-A2 was co-expressed with the MRL/lpr or C3H/lpr AHR in COS-7, FICZ treatment decreased EC50 to about threefold in both AHR genotypes, compared with EC50 in AHR alone. Similarly, hnRNP-A2b co-expression also lowered the FICZ-EC50 values. In TCDD-treated COS-7, responses depended on the AHR genotype; while no change in TCDD-EC50 was observed for C3H/lpr AHR when hnRNP-A2 was co-expressed, the value was reduced to nearly tenfold for MRL/lpr AHR. Co-transfection with hnRNP-A2b attenuated the AHR sensitivity to TCDD. In conclusion, the hnRNP-A2/B1 interacting with AHR may be a modulator of the AHR ligand sensitivity.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
DL-二硫代苏糖醇 溶液, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-二硫代苏糖醇 溶液, 1 M in H2O
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
二甲基亚砜, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
三(叔丁氧基)硅烷醇, 99.999%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
三(叔丁氧基)硅烷醇, packaged for use in deposition systems
Sigma-Aldrich
二甲基亚砜, ≥99.6%, ReagentPlus®