Merck
  • 主页
  • 查找结果
  • Comprehensive cytotoxic evaluation of morin, a bioflavonoid against verapamil on rat gastrointestinal epithelium for novel pharmaceutical application involving P-glycoprotein inhibition.
Comprehensive cytotoxic evaluation of morin, a bioflavonoid against verapamil on rat gastrointestinal epithelium for novel pharmaceutical application involving P-glycoprotein inhibition.
The Journal of pharmacy and pharmacology (2015-04-14)
Gurunath Surampalli, Basavaraj K Nanjwade, Paragouda A Patil
摘要
In this study, a comprehensive and comparative cytotoxic evaluation of morin against verapamil on rat intestinal epithelium as P-gp inhibitors through in-vitro gastrointestinal short-term toxicity assays involving permeability studies for safety evaluation was investigated. In this study, the effect of morin (1 mM or 10 mM) or verapamil (1 mM or 10 mM) or sodium deoxycholate (10 mM) was investigated on intestinal epithelium and isolated brush border membrane using biomarker assays. Cytotoxicity was determined using 3-(4,5-dimethylthiazolyl-2)-2,5-diphenyltetrazolium bromide (MTT) assay. The nutrients transport was assessed using everted sacs studies. Paracellular permeability was measured using Lucifer yellow, followed by morphometric analysis of intestinal sacs. Our results indicated that morin was effective in maintaining cell viability with no significant changes (P > 0.05) in the activity of intestinal brush border markers, membrane integrity and morphometric analysis as compared with control. On the contrary, dramatic (P < 0.01) changes were noticed in the release of membrane markers, cell viability and surface characteristics of intestinal segments when treated with verapamil or sodium deoxycholate as compared with control or morin. Our findings confirm that morin is non-toxic to rat intestinal epithelium against verapamil demonstrating the potential use of bioflavonoid as safe and novel pharmaceutical adjuvant as P-gp inhibitor.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
碳酸氢钠, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
氯化镁 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
DL-二硫代苏糖醇 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化镁, anhydrous, ≥98%
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
氯化钙 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
氯化钾, for molecular biology, ≥99.0%
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Supelco
DL-二硫代苏糖醇 溶液, 1 M in H2O
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钾, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
L-组氨酸, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
碳酸钠, BioXtra, ≥99.0%
Sigma-Aldrich
氯化钾, BioXtra, ≥99.0%
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%