Merck
  • 主页
  • 查找结果
  • A Simple and Green Route for Room-Temperature Synthesis of Gold Nanoparticles and Selective Colorimetric Detection of Cysteine.
A Simple and Green Route for Room-Temperature Synthesis of Gold Nanoparticles and Selective Colorimetric Detection of Cysteine.
Journal of food science (2015-08-05)
Pelin Onsekizoglu Bagci, Yi-Cheng Wang, Sundaram Gunasekaran
摘要
Gold nanoparticles (AuNPs) were synthesized at room temperature following a simple, rapid, and green route using fresh-squeezed apple juice as a reducing reagent. The optimal AuNPs, based on the particle color, stability, and color change suitable for colorimetric detection of cysteine (Cys), are synthesized using 5 mL of 10% apple juice, 1 mL of 10 mM gold precursor solution, and 1 mL of 0.1 M NaOH. Under this set of parameters, the AuNPs are synthesized within 30 min at room temperature. The average size (11.1 ± 3.2 nm) and ζ potential (-36.5 mV) of the AuNPs synthesized were similar to those of AuNPs prepared via the conventional citrate-reduction method. In the presence of Cys, unlike with any other amino acid, the AuNPs aggregated, possibly due to the gold-sulfur covalent interaction, yielding red-to-purple color change of the sample solution. The red-shift of the localized surface plasmon resonance peak of the AuNPs responsible for the color change was recorded by UV-vis spectrometer. The effect of other potential interferents such as glucose, ascorbic acid, K(+) , Na(+) , Ca(2+) , Zn(2+) , Ag(+) , Ni(2+) , Cu(2+) , Co(2+) , and Hg(2+) were also examined. The results show that AuNPs can be used to selectively detect and measure Cys with a linear dependency in the range of 2 to 100 μM and a limit of detection (signal-to-noise ratio > 3) of 50 nM. The results suggest that the green-synthesized AuNPs are useful for simple, rapid, and sensitive colorimetric detection of Cys, which is an essential amino acid in food and biological systems.
材料
货号
品牌
产品描述
Sigma-Aldrich
氢氧化钠, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
氯化钾, ACS reagent, 99.0-100.5%
Sigma-Aldrich
D -(+)-葡萄糖, ≥99.5% (GC)
Sigma-Aldrich
氢氧化钠, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
D -(+)-葡萄糖, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氢氧化钠 溶液, 50% in H2O
Sigma-Aldrich
L -抗坏血酸, 99%
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
氯化钙 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
氯化钾, for molecular biology, ≥99.0%
Sigma-Aldrich
氯化钙 二水合物, ACS reagent, ≥99%
Sigma-Aldrich
葡萄糖, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
色氨酸 -L, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
氢氧化钠, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
L-精氨酸, reagent grade, ≥98%
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-精氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-半胱氨酸, 97%
Sigma-Aldrich
L -抗坏血酸, powder, suitable for cell culture, γ-irradiated
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
L-脯氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
D -(+)-葡萄糖, ACS reagent