Merck
  • 主页
  • 查找结果
  • Expression Analyses Revealed Thymic Stromal Co-Transporter/Slc46A2 Is in Stem Cell Populations and Is a Putative Tumor Suppressor.
Expression Analyses Revealed Thymic Stromal Co-Transporter/Slc46A2 Is in Stem Cell Populations and Is a Putative Tumor Suppressor.
Molecules and cells (2015-05-28)
Ki Yeon Kim, Gwanghee Lee, Minsang Yoon, Eun Hye Cho, Chan-Sik Park, Moon Gyo Kim
摘要
By combining conventional single cell analysis with flow cytometry and public database searches with bioinformatics tools, we extended the expression profiling of thymic stromal cotransporter (TSCOT), Slc46A2/Ly110, that was shown to be expressed in bipotent precursor and cortical thymic epithelial cells. Genome scale analysis verified TSCOT expression in thymic tissue- and cell type- specific fashion and is also expressed in some other epithelial tissues including skin and lung. Coexpression profiling with genes, Foxn1 and Hoxa3, revealed the role of TSCOT during the organogenesis. TSCOT expression was detected in all thymic epithelial cells (TECs), but not in the CD31(+) endothelial cell lineage in fetal thymus. In addition, ABC transporter-dependent side population and Sca-1(+) fetal TEC populations both contain TSCOT-expressing cells, indicating TEC stem cells express TSCOT. TSCOT expression was identified as early as in differentiating embryonic stem cells. TSCOT expression is not under the control of Foxn1 since TSCOT is present in the thymic rudiment of nude mice. By searching variations in the expression levels, TSCOT is positively associated with Grhl3 and Irf6. Cytokines such as IL1b, IL22 and IL24 are the potential regulators of the TSCOT expression. Surprisingly, we found TSCOT expression in the lung is diminished in lung cancers, suggesting TSCOT may be involved in the suppression of lung tumor development. Based on these results, a model for TEC differentiation from the stem cells was proposed in context of multiple epithelial organ formation.
材料
货号
品牌
产品描述
Sigma-Aldrich
N,N-二甲基甲酰胺, anhydrous, 99.8%
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
氯化镁 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化镁, anhydrous, ≥98%
Sigma-Aldrich
戊二醛 溶液, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
戊二醛 溶液, Grade II, 25% in H2O
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
戊二醛 溶液, 50 wt. % in H2O
Sigma-Aldrich
N,N-二甲基甲酰胺, for molecular biology, ≥99%
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
戊二醛 溶液, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
戊二醛 溶液, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
戊二醛 溶液, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
钾, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
氯化镁, powder, <200 μm
Sigma-Aldrich
戊二醛 溶液, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氢化钾, 30 wt % dispersion in mineral oil
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, 2 M in H2O