Merck
  • 主页
  • 查找结果
  • Screening of Non- Saccharomyces cerevisiae Strains for Tolerance to Formic Acid in Bioethanol Fermentation.
Screening of Non- Saccharomyces cerevisiae Strains for Tolerance to Formic Acid in Bioethanol Fermentation.
PloS one (2015-08-19)
Cyprian E Oshoma, Darren Greetham, Edward J Louis, Katherine A Smart, Trevor G Phister, Chris Powell, Chenyu Du
摘要
Formic acid is one of the major inhibitory compounds present in hydrolysates derived from lignocellulosic materials, the presence of which can significantly hamper the efficiency of converting available sugars into bioethanol. This study investigated the potential for screening formic acid tolerance in non-Saccharomyces cerevisiae yeast strains, which could be used for the development of advanced generation bioethanol processes. Spot plate and phenotypic microarray methods were used to screen the formic acid tolerance of 7 non-Saccharomyces cerevisiae yeasts. S. kudriavzeii IFO1802 and S. arboricolus 2.3319 displayed a higher formic acid tolerance when compared to other strains in the study. Strain S. arboricolus 2.3319 was selected for further investigation due to its genetic variability among the Saccharomyces species as related to Saccharomyces cerevisiae and availability of two sibling strains: S. arboricolus 2.3317 and 2.3318 in the lab. The tolerance of S. arboricolus strains (2.3317, 2.3318 and 2.3319) to formic acid was further investigated by lab-scale fermentation analysis, and compared with S. cerevisiae NCYC2592. S. arboricolus 2.3319 demonstrated improved formic acid tolerance and a similar bioethanol synthesis capacity to S. cerevisiae NCYC2592, while S. arboricolus 2.3317 and 2.3318 exhibited an overall inferior performance. Metabolite analysis indicated that S. arboricolus strain 2.3319 accumulated comparatively high concentrations of glycerol and glycogen, which may have contributed to its ability to tolerate high levels of formic acid.
材料
货号
品牌
产品描述
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
色氨酸 -L, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
盐酸, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
盐酸 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-组氨酸, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L -赖氨酸, ≥98% (TLC)
Sigma-Aldrich
L-亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-亮氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-甲硫氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
腺嘌呤, ≥99%
SAFC
L-甲硫氨酸
Sigma-Aldrich
色氨酸 -L, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
盐酸 溶液, volumetric, 0.1 M HCl (0.1N), endotoxin free
SAFC
L-组氨酸
Sigma-Aldrich
色氨酸 -L, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
腺嘌呤, BioReagent, suitable for cell culture
Sigma-Aldrich
L-组氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
盐酸 溶液, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
盐酸 溶液, 32 wt. % in H2O, FCC
Sigma-Aldrich
L-甲硫氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L -赖氨酸, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-亮氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
氯化氢 溶液, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
L-亮氨酸, 99%, FG
Sigma-Aldrich
腺嘌呤, BioReagent, suitable for plant cell culture, ≥99%