Merck
  • 主页
  • 查找结果
  • A conserved role for Notch signaling in priming the cellular response to Shh through ciliary localisation of the key Shh transducer Smo.
A conserved role for Notch signaling in priming the cellular response to Shh through ciliary localisation of the key Shh transducer Smo.
Development (Cambridge, England) (2015-05-23)
Magdalena Stasiulewicz, Shona D Gray, Ioanna Mastromina, Joana C Silva, Mia Björklund, Philip A Seymour, David Booth, Calum Thompson, Richard J Green, Emma A Hall, Palle Serup, J Kim Dale
摘要
Notochord-derived Sonic Hedgehog (Shh) is essential for dorsoventral patterning of the overlying neural tube. Increasing concentration and duration of Shh signal induces progenitors to acquire progressively more ventral fates. We show that Notch signalling augments the response of neuroepithelial cells to Shh, leading to the induction of higher expression levels of the Shh target gene Ptch1 and subsequently induction of more ventral cell fates. Furthermore, we demonstrate that activated Notch1 leads to pronounced accumulation of Smoothened (Smo) within primary cilia and elevated levels of full-length Gli3. Finally, we show that Notch activity promotes longer primary cilia both in vitro and in vivo. Strikingly, these Notch-regulated effects are Shh independent. These data identify Notch signalling as a novel modulator of Shh signalling that acts mechanistically via regulation of ciliary localisation of key components of its transduction machinery.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
泰莫西芬, ≥99%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
戊二醛 溶液, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
戊二醛 溶液, Grade II, 25% in H2O
Sigma-Aldrich
戊二醛 溶液, 50 wt. % in H2O
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
抗乙酰化微管蛋白抗体,小鼠单克隆 小鼠抗, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
戊二醛 溶液, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
戊二醛 溶液, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
戊二醛 溶液, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
戊二醛 溶液, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
甲醛 溶液, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
戊二醛 溶液, 50 wt. % in H2O, FCC
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
甲醛-12C 溶液, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
二甲基亚砜, ≥99.6%, ReagentPlus®