  • Search Results
  • NMR structure and function of Helicoverpa armigera sterol carrier protein-2, an important insecticidal target from the cotton bollworm.
NMR structure and function of Helicoverpa armigera sterol carrier protein-2, an important insecticidal target from the cotton bollworm.
Scientific reports (2015-12-15)
Haihao Ma, Yuemin Ma, Xuehui Liu, David H Dyer, Pingyong Xu, Kaiyu Liu, Que Lan, Huazhu Hong, Jianxin Peng, Rong Peng
ABSTRACT
The cotton bollworm, Helicoverpa armigera, has developed strong resistance to many insecticides. Sterol Carrier Protein-2 (SCP-2) is an important non-specific lipid transfer protein in insects and appears to be a potential new target. In order to elucidate the structure and function of Helicoverpa armigera SCP-2 (HaSCP-2), NMR spectroscopy, docking simulations, mutagenesis and bioassays were performed. HaSCP-2 composed of five α-helices and four stranded β-sheets. The folds of α-helices and β-sheets interacted together to form a hydrophobic cavity with putative entrance and exit openings, which served as a tunnel for accommodating and transporting of lipids. Several sterols and fatty acids could interact with HaSCP-2 via important hydrophobic sites, which could be potential targets for insecticides. Mutagenesis experiments indicated Y51, F53, F89, F110, I117 and Q131 may be the key functional sites. HaSCP-2 showed high cholesterol binding activity and SCP-2 inhibitors (SCPIs) could inhibit the biological activity of HaSCP-2. SCPI-treated larvae at young stage showed a significant decrease of cholesterol uptake in vivo. Our study describes for the first time a NMR structure of SCP-2 in lepidopteran H. armigera and reveals its important function in cholesterol uptake, which facilitates the screening of effective insecticides targeting the insect cholesterol metabolism.
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
胆固醇, Sigma Grade, ≥99%
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
棕榈酸, ≥99%
Sigma-Aldrich
胆固醇, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SAFC
胆固醇、植物的, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
SyntheChol ® NS0 补充, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
胆固醇, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
棕榈酸, BioXtra, ≥99%
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
二喹啉甲酸 二钠盐 水合物, ≥98% (HPLC)
Sigma-Aldrich
棕榈酸, ≥95%, FCC, FG
Supelco
胆固醇 溶液, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
氯化钠, tablet
Sigma-Aldrich
棕榈酸, natural, 98%, FG
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
棕榈酸, ≥98% palmitic acid basis (GC)
SAFC
胆固醇, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
氯化钠, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.99% trace metals basis