  • Search Results
  • Methyl jasmonate stimulates biosynthesis of 2-phenylethylamine, phenylacetic acid and 2-phenylethanol in seedlings of common buckwheat.
Methyl jasmonate stimulates biosynthesis of 2-phenylethylamine, phenylacetic acid and 2-phenylethanol in seedlings of common buckwheat.
Acta biochimica Polonica (2015-04-10)
Marcin Horbowicz, Wiesław Wiczkowski, Tomasz Sawicki, Dorota Szawara-Nowak, Hubert Sytykiewicz, Joanna Mitrus
ABSTRACT
Methyl jasmonate has a strong effect on secondary metabolizm in plants, by stimulating the biosynthesis a number of phenolic compounds and alkaloids. Common buckwheat (Fagopyrum esculentum Moench) is an important source of biologically active compounds. This research focuses on the detection and quantification of 2-phenylethylamine and its possible metabolites in the cotyledons, hypocotyl and roots of common buckwheat seedlings treated with methyl jasmonate. In cotyledons of buckwheat sprouts, only traces of 2-phenylethylamine were found, while in the hypocotyl and roots its concentration was about 150 and 1000-times higher, respectively. Treatment with methyl jasmonate resulted in a 4-fold increase of the 2-phenylethylamine level in the cotyledons of 7-day buckwheat seedlings, and an 11-fold and 5-fold increase in hypocotyl and roots, respectively. Methyl jasmonate treatment led also to about 4-fold increase of phenylacetic acid content in all examined seedling organs, but did not affect the 2-phenylethanol level in cotyledons, and slightly enhanced in hypocotyl and roots. It has been suggested that 2-phenylethylamine is a substrate for the biosynthesis of phenylacetic acid and 2-phenylethanol, as well as cinnamoyl 2-phenethylamide. In organs of buckwheat seedling treated with methyl jasmonate, higher amounts of aromatic amino acid transaminase mRNA were found. The enzyme can be involved in the synthesis of phenylpyruvic acid, but the presence of this compound could not be confirmed in any of the examined organs of common buckwheat seedling.
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Sigma-Aldrich
2-苯乙醇, ≥99.0% (GC)
Sigma-Aldrich
苯乙醛, ≥90%
Sigma-Aldrich
苯乙醇, natural, ≥99%, FCC, FG
Sigma-Aldrich
苯乙醇, ≥99%, FCC, FG
Sigma-Aldrich
苯乙醛, ≥95%, FCC, FG
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-苯基丙氨酸, 99%
Sigma-Aldrich
DL-酪氨酸, 99%
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC
SAFC
L-苯基丙氨酸