Merck
  • 主页
  • 查找结果
  • Highly Efficient Photocatalytic Hydrogen Production of Flower-like Cadmium Sulfide Decorated by Histidine.
Highly Efficient Photocatalytic Hydrogen Production of Flower-like Cadmium Sulfide Decorated by Histidine.
Scientific reports (2015-09-05)
Qizhao Wang, Juhong Lian, Jiajia Li, Rongfang Wang, Haohao Huang, Bitao Su, Ziqiang Lei
摘要
Morphology-controlled synthesis of CdS can significantly enhance the efficiency of its photocatalytic hydrogen production. In this study, a novel three-dimensional (3D) flower-like CdS is synthesized via a facile template-free hydrothermal process using Cd(NO3)2•4H2O and thiourea as precursors and L-Histidine as a chelating agent. The morphology, crystal phase, and photoelectrochemical performance of the flower-like CdS and pure CdS nanocrystals are carefully investigated via various characterizations. Superior photocatalytic activity relative to that of pure CdS is observed on the flower-like CdS photocatalyst under visible light irradiation, which is nearly 13 times of pure CdS. On the basis of the results from SEM studies and our analysis, a growth mechanism of flower-like CdS is proposed by capturing the shape evolution. The imidazole ring of L-Histidine captures the Cd ions from the solution, and prevents the growth of the CdS nanoparticles. Furthermore, the photocatalytic contrast experiments illustrate that the as-synthesized flower-like CdS with L-Histidine is more stable than CdS without L-Histidine in the hydrogen generation.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-组氨酸, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
硫脲, ACS reagent, ≥99.0%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
硫脲, ReagentPlus®, ≥99.0%
SAFC
L-组氨酸
Sigma-Aldrich
L-组氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
亚硫酸钠, BioXtra, ≥98%
Sigma-Aldrich
亚硫酸钠, BioUltra, anhydrous, ≥98% (RT)
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
硫酸钠, ≥99.99% trace metals basis
Sigma-Aldrich
硫酸钠, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
硫酸钠, BioXtra, ≥99.0%
Sigma-Aldrich
硫酸钠, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
80% v/v 乙醇固定液, suitable for fixing solution (blood films)
Sigma-Aldrich
纯乙醇, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
纯乙醇, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
纯乙醇, 160 proof, Excise Tax-free, Permit for use required