• 主页
  • 查找结果
  • Effect of granule size on the properties of lotus rhizome C-type starch.
Effect of granule size on the properties of lotus rhizome C-type starch.
Carbohydrate polymers (2015-10-03)
Lingshang Lin, Jun Huang, Lingxiao Zhao, Juan Wang, Zhifeng Wang, Cunxu Wei
摘要
Lotus rhizome C-type starch was separated into different size fractions. Starch morphologies changed from irregular to elongated, ellipsoid, oval, and spherical with decreasing granule size. The small- and very-small-sized fractions had a centric hilum, and the other size fractions had an eccentric hilum. The different size fractions all showed C-type crystallinity, pseudoplasticity and shear-thinning rheological properties. The range of amylose content was 25.6 to 26.6%, that of relative crystallinity was 23.9 to 25.8%, that of swelling power was 29.0 to 31.4 g/g, and that of gelatinization enthalpy was 12.4 to 14.2J/g. The very-small-sized fraction had a significantly lower short-range ordered degree and flow behavior index and higher scattering peak intensity, water solubility, gelatinization peak temperature, gelatinization conclusion temperature, consistency coefficient, hydrolysis degrees, and digestion rate than the large-sized fraction. Granule size significantly positively influenced short-range ordered structure and swelling power and negatively influenced scattering peak intensity, water solubility, hydrolysis and digestion of starch (p<0.01).
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
纯乙醇, 200 proof, for molecular biology
Sigma-Aldrich
淀粉葡萄糖苷酶 来源于黑曲霉, ≥260 U/mL, aqueous solution
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
乙醇, anhydrous, denatured
Sigma-Aldrich
叠氮化钠, ReagentPlus®, ≥99.5%
Sigma-Aldrich
乙醇, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
纯乙醇, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
纯乙醇, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
磷酸钠, 96%
Sigma-Aldrich
氯化钙 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
α-淀粉酶 来源于猪胰腺, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
叠氮化钠, BioXtra
Sigma-Aldrich
氯化钙, meets USP testing specifications
Sigma-Aldrich
叠氮化钠, BioUltra, ≥99.5% (T)
Sigma-Aldrich
纯乙醇, 200 proof, meets USP testing specifications
Sigma-Aldrich
氯化钙, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
纯乙醇, 190 proof, for molecular biology
Sigma-Aldrich
氯化钙, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
叠氮化钠, purum p.a., ≥99.0% (T)
Sigma-Aldrich
乙醇, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
乙醇, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
氯化钙, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Ethanol 溶液, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.999% trace metals basis
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
乙醇, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
乙醇, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
乙醇, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)