  • Search Results
  • Taurine enhances excitability of mouse cochlear neural stem cells by selectively promoting differentiation of glutamatergic neurons over GABAergic neurons.
Taurine enhances excitability of mouse cochlear neural stem cells by selectively promoting differentiation of glutamatergic neurons over GABAergic neurons.
Neurochemical research (2015-03-03)
Qin Wang, Gang-Hua Zhu, Ding-Hua Xie, Wei-Jing Wu, Peng Hu
ABSTRACT
Taurine is a sulfur-containing amino acid present in high concentrations in mammalian tissues, and has been implicated in several processes involving brain development and neurotransmission. However, the role of taurine in inner ear neural development is still largely unknown. Here we report that taurine enhanced the viability and proliferation of in vitro mouse cochlear neural stem cell culture, as well as improved neurite outgrowth. Moreover, prolonged taurine treatment also increased the neural electrical activity by escalating changes of intracellular calcium concentration, the number of spontaneous Ca(2+) oscillations in cells, and the frequencies of Ca(2+) spikes. Most importantly, we found that this escalated neural excitability by taurine was due to combined effect of increase in the population of excitatory glutamatergic neuron and decrease in inhibitory GABAergic neuron population. This is the first report on the effect of taurine to selectively promote neural stem cell differentiation by altering neuron type commitment. Our study has supported the potential of taurine as treatment against hearing loss caused by neuron degeneration, or even as an agent to improve sensitivity of hearing by increasing overall excitability of auditory nervous system.
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
视黄酸, ≥98% (HPLC), powder
Sigma-Aldrich
氯化镁 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化镁, anhydrous, ≥98%
Sigma-Aldrich
噻唑蓝, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
噻唑蓝, 98%
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
氯化镁, powder, <200 μm
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
乙二胺四乙酸, 99.995% trace metals basis
Sigma-Aldrich
牛磺酸, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
二喹啉甲酸 二钠盐 水合物, ≥98% (HPLC)
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
氯化镁, BioReagent, suitable for insect cell culture, ≥97.0%