• 主页
  • 查找结果
  • Effect of olfactory manganese exposure on anxiety-related behavior in a mouse model of iron overload hemochromatosis.
Effect of olfactory manganese exposure on anxiety-related behavior in a mouse model of iron overload hemochromatosis.
Environmental toxicology and pharmacology (2015-07-21)
Qi Ye, Jonghan Kim
摘要
Manganese in excess promotes unstable emotional behavior. Our previous study showed that olfactory manganese uptake into the brain is altered in Hfe(-/-) mice, a model of iron overload hemochromatosis, suggesting that Hfe deficiency could modify the neurotoxicity of airborne manganese. We determined anxiety-related behavior and monoaminergic protein expression after repeated intranasal instillation of MnCl2 to Hfe(-/-) mice. Compared with manganese-instilled wild-type mice, Hfe(-/-) mice showed decreased manganese accumulation in the cerebellum. Hfe(-/-) mice also exhibited increased anxiety with decreased exploratory activity and elevated dopamine D1 receptor and norepinephrine transporter in the striatum. Moreover, Hfe deficiency attenuated manganese-associated impulsivity and modified the effect of manganese on the expression of tyrosine hydroxylase, vesicular monoamine transporter and serotonin transporter. Together, our data indicate that loss of HFE function alters manganese-associated emotional behavior and further suggest that HFE could be a potential molecular target to alleviate affective disorders induced by manganese inhalation.
材料
货号
品牌
产品描述
Sigma-Aldrich
硝酸, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
硝酸, ACS reagent, ≥90.0%
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%