• 主页
  • 查找结果
  • Melatonin protects hepatocytes against bile acid-induced mitochondrial oxidative stress via the AMPK-SIRT3-SOD2 pathway.
Melatonin protects hepatocytes against bile acid-induced mitochondrial oxidative stress via the AMPK-SIRT3-SOD2 pathway.
Free radical research (2015-06-30)
Y Chen, W Qing, M Sun, L Lv, D Guo, Y Jiang
摘要
Mitochondrial oxidative damage is hypothesized to contribute to the pathogenesis of chronic cholestatic liver diseases. Melatonin, an indolamine synthesized in the pineal gland, shows a wide range of physiological functions, and is under clinical investigation for expanded applications. Melatonin has demonstrated efficient protective effects against various types of oxidative damage in the liver system. This study investigates the protective effects of melatonin pretreatment on glycochenodeoxycholic acid (GCDCA)-induced hepatotoxicity and elucidates the potential mechanism of melatonin-mediated protection. Melatonin markedly decreased mitochondrial ROS (mROS) production in L02 cells treated with 100 μM GCDCA, and inhibited GCDCA-stimulated cytotoxicity. Notably, melatonin exerted its hepatoprotective effects by upregulating sirtuin 3 (SIRT3) activity and its expression level, thus regulating superoxide dismutase 2 (SOD2) acetylation and inhibiting the production of mROS induced by GCDCA. Moreover, siRNA targeting SIRT3 blocked the melatonin-mediated elevation in mitochondrial function by inhibiting SIRT3/SOD2 signaling. Importantly, melatonin-activated SIRT3 activity was completely abolished by AMP-activated, alpha 1 catalytic subunit (AMPK) siRNA transfection. Similar results were obtained in rat with bile duct ligation or BDL. In summary, our findings indicate that melatonin is a novel hepatoprotective small molecule that functions by elevating SIRT3, stimulating SOD2 activity, and suppressing mitochondrial oxidative stress at least through AMPK, and that SIRT3 may be of therapeutic value in liver cell protection for GCDCA-induced hepatotoxicity.
材料
货号
品牌
产品描述
Sigma-Aldrich
褪黑素, powder, ≥98% (TLC)
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
JC-1, solid
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%