• 主页
  • 查找结果
  • Effects of inhaled CO administration on acute lung injury in baboons with pneumococcal pneumonia.
Effects of inhaled CO administration on acute lung injury in baboons with pneumococcal pneumonia.
American journal of physiology. Lung cellular and molecular physiology (2015-09-01)
Laura E Fredenburgh, Bryan D Kraft, Dean R Hess, R Scott Harris, Monroe A Wolf, Hagir B Suliman, Victor L Roggli, John D Davies, Tilo Winkler, Alex Stenzler, Rebecca M Baron, B Taylor Thompson, Augustine M Choi, Karen E Welty-Wolf, Claude A Piantadosi
摘要
Inhaled carbon monoxide (CO) gas has therapeutic potential for patients with acute respiratory distress syndrome if a safe, evidence-based dosing strategy and a ventilator-compatible CO delivery system can be developed. In this study, we used a clinically relevant baboon model of Streptococcus pneumoniae pneumonia to 1) test a novel, ventilator-compatible CO delivery system; 2) establish a safe and effective CO dosing regimen; and 3) investigate the local and systemic effects of CO therapy on inflammation and acute lung injury (ALI). Animals were inoculated with S. pneumoniae (10(8)-10(9) CFU) (n = 14) or saline vehicle (n = 5); in a subset with pneumonia (n = 5), we administered low-dose, inhaled CO gas (100-300 ppm × 60-90 min) at 0, 6, 24, and/or 48 h postinoculation and serially measured blood carboxyhemoglobin (COHb) levels. We found that CO inhalation at 200 ppm for 60 min is well tolerated and achieves a COHb of 6-8% with ambient CO levels ≤ 1 ppm. The COHb level measured at 20 min predicted the 60-min COHb level by the Coburn-Forster-Kane equation with high accuracy. Animals given inhaled CO + antibiotics displayed significantly less ALI at 8 days postinoculation compared with antibiotics alone. Inhaled CO was associated with activation of mitochondrial biogenesis in the lung and with augmentation of renal antioxidative programs. These data support the feasibility of safely delivering inhaled CO gas during mechanical ventilation and provide preliminary evidence that CO may accelerate the resolution of ALI in a clinically relevant nonhuman primate pneumonia model.
材料
货号
品牌
产品描述
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠 溶液, 5 M
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
鲁米诺, 97%
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
DL-甘油醛-3-磷酸 溶液, 45-55 mg/mL in H2O
Sigma-Aldrich
鲁米诺, ≥97% (HPLC)
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
氯化钠, tablet
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.99% trace metals basis
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%