  • Search Results
  • Phenylalanine Blocks Defects Induced in Gel Lipid Membranes by Osmotic Stress.
Phenylalanine Blocks Defects Induced in Gel Lipid Membranes by Osmotic Stress.
The journal of physical chemistry. B (2015-07-29)
A C Cutró, E A Disalvo
ABSTRACT
We study the binding of phenylalanine (Phe) with dipalmitoylphosphocholine (DPPC) vesicles in gel (25 °C) and in liquid crystalline states (50 °C) and in gel large unilamellar vesicles (LUVs) subjected to osmotic dehydration with merocyanine (MC 540) as a fluorescent surface membrane marker. Phe does not produce significant changes in MC 540 monomer concentration in DPPC LUVs at 50 °C. In contrast, it significantly decreases the monomer adsorption in defects present in DPPC LUVs at 25 °C. When DPPC LUVs were subjected to hypertonic stress, dehydration caused more defects, and in this case phenylalanine is also able to block such defects.
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
L-苯基丙氨酸, 99%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
份菁 540, Dye content 90 %
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, Vetec, reagent grade, 99.5%
SAFC
L-苯基丙氨酸