  • Search Results
  • Clinical performance and safety of adapters for intradermal delivery with conventional and autodisable syringes.
Clinical performance and safety of adapters for intradermal delivery with conventional and autodisable syringes.
Vaccine (2015-05-13)
Izrail Tsals, Courtney Jarrahian, Fred E Snyder, Laura Saganic, Eugene Saxon, Darin Zehrung, Glen Zimmerman, Mark Papania, Leslie Klaff
ABSTRACT
Although the number of vaccines and diagnostic tests currently delivered intradermally is limited, this route of administration offers potential advantages due to the high concentration of antigen-presenting cells in the skin. One factor which may in part be limiting development and use of intradermal (ID) administration is concern about the ease and reliability of the needle and syringe-based Mantoux technique. A phase I clinical study was conducted to evaluate two ID adapters that have been developed as injection-delivery aids to increase the safety, simplicity, and reliability of ID injection: a prototype autodisable, intradermal (ADID) adapter for autodisable (AD) syringes, and a marketed side-merge adapter (SMA). Thirty healthy adult volunteers each received six injections of 0.1 mL of sterile saline solution. Each adapter was used to give injections into the upper deltoid, forearm, and suprascapular regions of each volunteer. The needle-bevel orientation during injection was random. Injection performance was determined by measuring wheal size and fluid leakage. Wheals were similar in size for the ADID adapter (mean 9.9 ± 0.17 mm) and SMA (mean 9.8 ± 0.15 mm). In all of the injections completed with the SMA, and 98% of those completed with the ADID, fluid leakage was less than 10% of the intended injection volume. Minor skin abrasions were the only adverse events. Based on self-reporting of pain, injections were well tolerated (mean pain score of 2 on a 0-10 scale). ID delivery using the SMA and ADID adapters appears safe and effective.
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
氯化钠, tablet
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
甲基丙烯酸丙酮缩甘油酯, 50 wt. % in dichloromethane, contains ~280 ppm 4-tert-butylcatechol as inhibitor
Sigma-Aldrich
氯化钠, AR, ≥99.9%
Sigma-Aldrich
氯化钠, ≥99%, AR grade
Sigma-Aldrich
氯化钠-35Cl, 99 atom % 35Cl