• 主页
  • 查找结果
  • High catalytic activity of oriented 2.0.0 copper(I) oxide grown on graphene film.
High catalytic activity of oriented 2.0.0 copper(I) oxide grown on graphene film.
Nature communications (2015-10-29)
Ana Primo, Ivan Esteve-Adell, Juan F Blandez, Amarajothi Dhakshinamoorthy, Mercedes Álvaro, Natalia Candu, Simona M Coman, Vasile I Parvulescu, Hermenegildo García
摘要
Metal oxide nanoparticles supported on graphene exhibit high catalytic activity for oxidation, reduction and coupling reactions. Here we show that pyrolysis at 900 °C under inert atmosphere of copper(II) nitrate embedded in chitosan films affords 1.1.1 facet-oriented copper nanoplatelets supported on few-layered graphene. Oriented (1.1.1) copper nanoplatelets on graphene undergo spontaneous oxidation to render oriented (2.0.0) copper(I) oxide nanoplatelets on few-layered graphene. These films containing oriented copper(I) oxide exhibit as catalyst turnover numbers that can be three orders of magnitude higher for the Ullmann-type coupling, dehydrogenative coupling of dimethylphenylsilane with n-butanol and C-N cross-coupling than those of analogous unoriented graphene-supported copper(I) oxide nanoplatelets.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
纯乙醇, 200 proof, for molecular biology
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
乙醇, anhydrous, denatured
Sigma-Aldrich
乙醇, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
纯乙醇, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
纯乙醇, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
纯乙醇, 200 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, for molecular biology
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
乙醇, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
乙醇, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Supelco
Ethanol 溶液, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, powder, <75 μm, 99%
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
乙醇, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis