• 主页
  • 查找结果
  • Sphingomonas panacis sp. nov., isolated from rhizosphere of rusty ginseng.
Sphingomonas panacis sp. nov., isolated from rhizosphere of rusty ginseng.
Antonie van Leeuwenhoek (2015-07-15)
Priyanka Singh, Yeon-Ju Kim, Van-An Hoang, Mohamed El-Agamy Farh, Deok-Chun Yang
摘要
The type strain DCY99(T) was isolated from soil collected from a ginseng field in Hwacheon, Republic of Korea. Strain DCY99(T) is Gram-negative, non-spore forming, motile, rod-shaped, and strictly aerobic. The bacteria grow optimally at 25-30 °C and pH 6.0-6.5. Phylogenetically, strain DCY99(T) is most closely related to Sphingomonas oligophenolica JCM 12082(T), followed by Sphingomonas asaccharolytica KCTC 2825(T), Sphingomonas mali KCTC 2826(T), Sphingomonas cynarae JCM17498(T), Sphingomonas pruni KCTC 2824(T), and Sphingomonas glacialis DSM 22294(T). The DNA-DNA relatedness between strain DCY99(T) and S. oligophenolica JCM 12082(T) was 15.6 ± 0.4 %, and the DNA G+C content of strain DCY99(T) was 64.4 mol%. An isoprenoid quinone was detected and identified as ubiquinone Q-10, and sym-homospermidine was identified as the major polyamine of DCY99(T). The major polar lipids were identified as sphingoglycolipid, diphosphatidylglycerol, phosphatidylethanolamine, phosphatidylglycerol, and phosphatidylcholine. C14:02OH, C16:0, and summed feature 8 (C18:1 ω7c:/C18:1 ω6c) were identified as the major fatty acids present in DCY99(T). The results of physiological and biochemical tests allowed strain DCY99(T) to be differentiated phenotypically from other recognized species belonging to the genus Sphingomonas. Therefore, it is suggested that the newly isolated organism represents a novel species, for which the name Sphingomonas panacis sp. nov. is proposed with the type strain designated as DCY99(T) (=JCM 30806(T) =KCTC 42347(T)).
材料
货号
品牌
产品描述
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
D -(+)-葡萄糖, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
甘油, ≥99.5%
Sigma-Aldrich
D -(+)-葡萄糖, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
葡萄糖, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钙 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
D -(+)-葡萄糖, ACS reagent
Sigma-Aldrich
甘油, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
精脒, ≥99% (GC)
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
D -(+)-葡萄糖, BioXtra, ≥99.5% (GC)
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
精脒, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
氯化钙, meets USP testing specifications
Sigma-Aldrich
氯化钙, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
氯化钙, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
1,4-二氨基丁烷, 99%
Sigma-Aldrich
硝酸银, 99.9999% trace metals basis
Sigma-Aldrich
甘油, meets USP testing specifications
Sigma-Aldrich
精脒, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
精胺, ≥97%
Sigma-Aldrich
甘油, BioXtra, ≥99% (GC)
Sigma-Aldrich
甘油 溶液, 83.5-89.5% (T)