  • Search Results
  • Bile Salts Modulate the Mucin-Activated Type VI Secretion System of Pandemic Vibrio cholerae.
Bile Salts Modulate the Mucin-Activated Type VI Secretion System of Pandemic Vibrio cholerae.
PLoS neglected tropical diseases (2015-09-01)
Verena Bachmann, Benjamin Kostiuk, Daniel Unterweger, Laura Diaz-Satizabal, Stephen Ogg, Stefan Pukatzki
ABSTRACT
The causative agent of cholera, Vibrio cholerae, regulates its diverse virulence factors to thrive in the human small intestine and environmental reservoirs. Among this pathogen's arsenal of virulence factors is the tightly regulated type VI secretion system (T6SS). This system acts as an inverted bacteriophage to inject toxins into competing bacteria and eukaryotic phagocytes. V. cholerae strains responsible for the current 7th pandemic activate their T6SS within the host. We established that T6SS-mediated competition occurs upon T6SS activation in the infant mouse, and that this system is functional under anaerobic conditions. When investigating the intestinal host factors mucins (a glycoprotein component of mucus) and bile for potential regulatory roles in controlling the T6SS, we discovered that once mucins activate the T6SS, bile acids can further modulate T6SS activity. Microbiota modify bile acids to inhibit T6SS-mediated killing of commensal bacteria. This interplay is a novel interaction between commensal bacteria, host factors, and the V. cholerae T6SS, showing an active host role in infection.
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
甘氨酸, suitable for electrophoresis, ≥99%
Sigma-Aldrich
甘氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
脱氧胆酸钠, ≥97% (titration)
Sigma-Aldrich
丙酸, ACS reagent, ≥99.5%
Sigma-Aldrich
脱氧胆酸钠, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
甘氨酸, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
1-丙醇, anhydrous, 99.7%
Sigma-Aldrich
甘氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
甘氨酸, ACS reagent, ≥98.5%
SAFC
甘氨酸
SAFC
脱氧胆酸钠
Sigma-Aldrich
甘氨酸, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
甘氨酸, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
乙酸异戊酯, ≥95%, FCC, FG
Sigma-Aldrich
牛磺酸, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
乙酸异戊酯, anhydrous, ≥99%
Sigma-Aldrich
丙酸, ≥99.5%, FCC, FG
Sigma-Aldrich
乙酸异戊酯, natural, ≥97%, FCC, FG
Sigma-Aldrich
1-丙醇, ≥99%, FG
Sigma-Aldrich
丙酸, natural, 99%, FG
Sigma-Aldrich
甘氨酸, BioXtra, ≥99% (titration)
Sigma-Aldrich
丙酸, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
牛磺酸, BioUltra, ≥99.5% (T)
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
甘氨酸, 99%, FCC
Sigma-Aldrich
丙酸, 99%