  • Search Results
  • Dexamethasone inhibits the differentiation of rat tendon stem cells into tenocytes by targeting the scleraxis gene.
Dexamethasone inhibits the differentiation of rat tendon stem cells into tenocytes by targeting the scleraxis gene.
The Journal of steroid biochemistry and molecular biology (2015-04-25)
Wan Chen, Hong Tang, Mei Zhou, Chao Hu, Jiqiang Zhang, Kanglai Tang
ABSTRACT
Glucocorticoid-induced tendon rupture is very common in clinical practice, and the overall outcome of surgical suture repair is rather poor. The mechanism remains unclear, and effective treatments are still lacking. In the present study, we investigated the effect of dexamethasone on the differentiation of rat tendon stem cells (TSCs) to tenocytes and the underlying molecular mechanisms and found that dexamethasone inhibits the differentiation of TSCs to tenocytes by analyzing the development of long, spindle-shaped cells and detecting the expression of tenocyte markers type I collagen and tenomodulin (TNMD) at both the mRNA and protein levels. We also discovered that after treatment with dexamethasone, the scleraxis expression level is downregulated in vitro and in human specimen. Chromatin immunoprecipitation (ChIP)-PCR showed that dexamethasone promotes glucocorticoid receptor interacted with the TGGAAGCC sequence located between -734 and -726 base pairs (bp) upstream of the start codon of the scleraxis gene. Furthermore, TSCs were transfected with scleraxis knockdown or overexpression plasmids, and the results indicated that scleraxis plays a pivotal role in the differentiation of TSCs to tenocytes. In conclusion, dexamethasone inhibits the differentiation of TSCs to tenocytes by inhibiting the scleraxis gene.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
十二烷基硫酸钠, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
十二烷基硫酸钠, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
十二烷基硫酸钠, ACS reagent, ≥99.0%
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
十二烷基硫酸钠 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
十二烷基硫酸钠, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
十二烷基硫酸钠, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
地塞米松, ≥98% (HPLC), powder
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
L-谷氨酰胺, ReagentPlus®, ≥99% (HPLC)
SAFC
L-谷氨酰胺
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
十二烷基硫酸钠, 92.5-100.5% based on total alkyl sulfate content basis