  • Search Results
  • Rapid multislice T1 mapping of mouse myocardium: Application to quantification of manganese uptake in α-Dystrobrevin knockout mice.
Rapid multislice T1 mapping of mouse myocardium: Application to quantification of manganese uptake in α-Dystrobrevin knockout mice.
Magnetic resonance in medicine (2014-11-20)
Kai Jiang, Wen Li, Wei Li, Sen Jiao, Laurie Castel, David R Van Wagoner, Xin Yu
ABSTRACT
The aim of this study was to develop a rapid, multislice cardiac T1 mapping method in mice and to apply the method to quantify manganese (Mn(2+)) uptake in a mouse model with altered Ca(2+) channel activity. An electrocardiography-triggered multislice saturation-recovery Look-Locker method was developed and validated both in vitro and in vivo. A two-dose study was performed to investigate the kinetics of T1 shortening, Mn(2+) relaxivity in myocardium, and the impact of Mn(2+) on cardiac function. The sensitivity of Mn(2+)-enhanced MRI in detecting subtle changes in altered Ca(2+) channel activity was evaluated in a mouse model with α-dystrobrevin knockout. Validation studies showed strong agreement between the current method and an established method. High Mn(2+) dose led to significantly accelerated T1 shortening. Heart rate decreased during Mn(2+) infusion, while ejection ratio increased slightly at the end of imaging protocol. No statistical difference in cardiac function was detected between the two dose groups. Mice with α-dystrobrevin knockout showed enhanced Mn(2+) uptake in vivo. In vitro patch-clamp study showed increased Ca(2+) channel activity. The saturation recovery method provides rapid T1 mapping in mouse hearts, which allowed sensitive detection of subtle changes in Mn(2+) uptake in α-dystrobrevin knockout mice.
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
硫酸镁, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
L -抗坏血酸, 99%
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L -抗坏血酸, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L -抗坏血酸, ACS reagent, ≥99%
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
L -抗坏血酸, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-谷氨酰胺, ReagentPlus®, ≥99% (HPLC)
SAFC
L-谷氨酰胺
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
硫酸镁, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
硫酸镁 溶液, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L -抗坏血酸, reagent grade, crystalline
Sigma-Aldrich
硫酸镁, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
L -抗坏血酸, analytical standard
Sigma-Aldrich
硫酸镁, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
L -抗坏血酸, meets USP testing specifications
Sigma-Aldrich
硫酸镁, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
牛磺酸, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
L -抗坏血酸, reagent grade
Sigma-Aldrich
硫酸镁, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
硫酸镁 溶液, BioUltra, for molecular biology