  • Search Results
  • Anti-adipogenic and antioxidant effects of the traditional Korean herbal formula Samchulgeonbi-tang: an in vitro study.
Anti-adipogenic and antioxidant effects of the traditional Korean herbal formula Samchulgeonbi-tang: an in vitro study.
International journal of clinical and experimental medicine (2015-08-27)
Sae-Rom Yoo, Chang-Seob Seo, Ohn-Soon Kim, Hyeun-Kyoo Shin, Soo-Jin Jeong
ABSTRACT
Here we report in vitro anti-adipogenic and antioxidant effects of Samchulgeonbi-tang (SCGBT), a traditional Korean herbal formula. 3T3-L1 preadipocytes were differentiated into adipocytes with or without SCGBT. After differentiation, we measured Oil Red O staining, glycerol-3-phosphate dehydrogenase (GPDH) activity and leptin production. In addition, its effect on scavenging activities of 2,2'-azinobis-(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) and 2,2'-diphenyl-1-picrylhydrazyl (DPPH) radicals in in vitro systems. In differentiated 3T3-L1 adipocytes, SCGBT significantly inhibited lipid accumulation and triglyceride production, and mediated inactivation of GPDH, a major enzyme in the process of adipogenesis. Consistent with this, SCGBT stimulation significantly decreased the amount of leptin in 3T3-L1 adipose cells. Furthermore, SCGBT enhanced the scavenging activities on ABTS and DPPH radicals. The generation of malondialdehyde (MDA) during low-density lipoprotein (LDL) oxidation was significantly reduced by SCGBT treatment. Of interest, SCGBT extract inhibited reactive oxygen species (ROS) generation in 3T3-L1 adipocytes. Overall, our findings suggest that SCGBT has the potential for anti-adipogenic activity and antioxidant properties.
Sigma-Aldrich
N-氯代丁二酰亚胺, 98%
Sigma-Aldrich
乙酸, natural, ≥99.5%, FG
Sigma-Aldrich
4,4′-二氨基二苯甲烷, ≥97.0% (GC)
Sigma-Aldrich
橙皮苷, ≥80%
Sigma-Aldrich
柚皮苷, ≥95% (HPLC)
Sigma-Aldrich
乙酸, ≥99.5%, FCC, FG
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
柚皮苷, ≥90%, from citrus fruit
Sigma-Aldrich
甘草酸 铵盐 来源于甘草根(甘草), ≥95.0% (NT)
Sigma-Aldrich
丙酮, natural, ≥97%
Sigma-Aldrich
乙酸, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
丙酮, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
二羟丙酮磷酸盐 二锂盐, ≥93% (enzymatic)
Sigma-Aldrich
甘草酸 铵盐 来源于甘草根(甘草), ≥70% (HPLC)
Sigma-Aldrich
Albiflorin, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
乙酸-12C2, 99.9 atom % 12C