• 主页
  • 查找结果
  • Cloning, expression, and characterization of a Coxiella burnetii Cu/Zn Superoxide dismutase.
Cloning, expression, and characterization of a Coxiella burnetii Cu/Zn Superoxide dismutase.
BMC microbiology (2015-05-13)
Robert E Brennan, Katalin Kiss, Rachael Baalman, James E Samuel
摘要
Periplasmically localized copper-zinc co-factored superoxide dismutase (SodC) enzymes have been identified in a wide range of Gram-negative bacteria and are proposed to protect bacteria from exogenously produced toxic oxygen radicals, which indicates the potential significance of a Coxiella burnetii SodC. Assays for SOD activity demonstrated that the cloned C. burnetii insert codes for a SOD that was active over a wide range of pH and inhibitable with 5 mM H2O2 and 1 mM sodium diethyldithiocarbamate, a characteristic of Cu/ZnSODs that distinguishes them from Fe or Mn SODs. The sodC was expressed by C. burnetii, has a molecular weight of approximately 18 kDa, which is consistent with the predicted molecular weight, and localized towards the periphery of C. burnetii. Over expression of the C. burnetii sodC in an E. coli sodC mutant restored resistance to H2O2 killing to wild type levels. We have demonstrated that C. burnetii does express a Cu/ZnSOD that is functional at low pH, appears to be excreted, and was able to restore H2O2 resistance in an E. coli sodC mutant. Taken together, these results indicate that the C. burnetii Cu/ZnSOD is a potentially important virulence factor.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
乙醇, anhydrous, denatured
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
2-巯基乙醇, ≥99.0%
Sigma-Aldrich
2-巯基乙醇, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
十二烷基硫酸钠, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
甘油, ≥99.5%
Sigma-Aldrich
十二烷基硫酸钠, ACS reagent, ≥99.0%
Sigma-Aldrich
十二烷基硫酸钠, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
十二烷基硫酸钠 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
甘油, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
十二烷基硫酸钠, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
十二烷基硫酸钠, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-巯基乙醇, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
十二烷基硫酸钠 溶液, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
鲁米诺, 97%
Sigma-Aldrich
十二烷基硫酸钠, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
甘油, meets USP testing specifications
Sigma-Aldrich
次黄嘌呤, ≥99.0%
Sigma-Aldrich
十二烷基硫酸钠, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
甘油, BioXtra, ≥99% (GC)
Sigma-Aldrich
甘油 溶液, 83.5-89.5% (T)
Supelco
十二烷基硫酸钠, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
次黄嘌呤, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
甘油, FCC, FG
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Sigma-Aldrich
甘油, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
黄嘌呤, ≥99%