• 主页
  • 查找结果
  • Photophysical properties of zinc phthalocyanine-uridine single walled carbon nanotube--conjugates.
Photophysical properties of zinc phthalocyanine-uridine single walled carbon nanotube--conjugates.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-05-13)
Racheal O Ogbodu, Edith K Amuhaya, Philani Mashazi, Tebello Nyokong
摘要
The photophysical properties of the conjugate of uridine and zinc mono carboxy phenoxy phthalocyanine (ZnMCPPc-uridine, 4) are reported in this work. The conjugate was also adsorbed onto single walled carbon nanotubes (ZnMCPPc-uridine-SWCNT, 5). The X-ray photoelectron spectroscopy of 4 showed three N 1s peaks while that of 5 showed four N 1s peak, a new peak at 399.4 eV of 5 was assigned to pyrrolidonic nitrogen, due to the interaction of the pyrrolic nitrogen of 4 with the oxygen moiety of SWCNT-COOH in 5. The triplet lifetime, triplet and singlet oxygen quantum yields of the zinc mono carboxy phenoxy phthalocyanine increased by over 40% in the presence of uridine. SWCNTs resulted in only a small quenching of the triplet state parameters of 4.
材料
货号
品牌
产品描述
Sigma-Aldrich
三乙基胺, ≥99.5%
Sigma-Aldrich
三乙基胺, ≥99%
Sigma-Aldrich
4-(二甲氨基)吡啶, ReagentPlus®, ≥99%
Sigma-Aldrich
三乙基胺, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
三乙基胺, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
尿核甙, ≥99%
Sigma-Aldrich
尿核甙, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
α-氰基-4-羟基肉桂酸, suitable for MALDI-TOF MS
Sigma-Aldrich
α-氰基-4-羟基肉桂酸, ≥98% (TLC), powder
Sigma-Aldrich
三乙基胺, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
尿核甙, BioUltra, ≥99%
Sigma-Aldrich
α-氰基-4-羟基肉桂酸, 99%
Sigma-Aldrich
三乙基胺, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
三乙基胺, ≥99.5%
Sigma-Aldrich
2-(二甲氨基)吡啶, 97%
Sigma-Aldrich
α-氰基-4-羟基肉桂酸, 97%
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
锗, chips, 99.999% trace metals basis
锗, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
锗, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
锗, disks, 20mm, thickness 1.0mm, single crystal, 100%
锗, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
锗, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
锗, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
锗, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
锗, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
锗, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
锗, rod, 6mm, diameter 6.0mm, single crystal, 100%
锗, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%