  • Search Results
  • Quantitative lipidomic analysis of plasma and plasma lipoproteins using MALDI-TOF mass spectrometry.
Quantitative lipidomic analysis of plasma and plasma lipoproteins using MALDI-TOF mass spectrometry.
Chemistry and physics of lipids (2015-05-26)
Jorge Serna, David García-Seisdedos, Alberto Alcázar, Miguel Ángel Lasunción, Rebeca Busto, Óscar Pastor
ABSTRACT
Knowledge of the plasma lipid composition is essential to clarify the specific roles of different lipid species in various pathophysiological processes. In this study, we developed an analytical strategy combining high-performance liquid chromatography with evaporative light scattering detection (HPLC-ELSD) and off-line coupling with matrix-assisted laser desorption/ionization with time-of-flight mass spectrometry (MALDI-TOF/MS) to determine the composition of plasma and major lipoproteins at two levels, lipid classes and lipid species. We confirmed the suitability of MALDI-TOF/MS as a quantitative measurement tool studying the linearity and repeatability for triglycerides (TG), phosphatidylethanolamine (PE) and phosphatidylcholine (PC). Moreover, data obtained with this method were correlated with other lipid classes and species measurements using currently available technologies. To establish the potential utility of our approach, human plasma very low density- (VLDL), low density- (LDL) and high density- (HDL) lipoproteins from 10 healthy donors were separated using ultracentrifugation, and compositions of nine lipid classes, cholesteryl esters (CE), TG, free cholesterol (FC), PE, phosphatidylinositol (PI), sulfatides (S), PC, lysophosphatidylcholine (LPC) and sphingomyelin (SM), analyzed. In total, 157 lipid species in plasma, 182 in LDL, 171 in HDL, and 148 in VLDL were quantified. The lipidomic profile was consistent with known differences in lipid classes, but also revealed unexpected differences in lipid species distribution of lipoproteins, particularly for LPC and SM. In summary, the methodology developed in this study constitutes a valid approach to determine the lipidomic composition of plasma and lipoproteins.
Sigma-Aldrich
二氯甲烷, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-丙醇, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
2-丙醇, anhydrous, 99.5%
Sigma-Aldrich
正己烷, anhydrous, 95%
Sigma-Aldrich
氯仿, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
乙酸钠, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
氯仿, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
异丙醇, ≥99.7%, FCC, FG
Sigma-Aldrich
甲酸铵, ≥99.995% trace metals basis
Sigma-Aldrich
2-丙醇, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-丙醇, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
甲酸铵 溶液, BioUltra, 10 M in H2O
Sigma-Aldrich
2,2,4-三甲基戊烷, anhydrous, 99.8%
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙酸钠, >99%, FCC, FG
Sigma-Aldrich
乙酸钠 溶液, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
乙酸钠, meets USP testing specifications, anhydrous
Sigma-Aldrich
乙酸钠, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
乙酸钠, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
乙酸钠, BioXtra, ≥99.0%
Sigma-Aldrich
丙酮, natural, ≥97%
Sigma-Aldrich
氯仿, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
丙酮, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
乙酸钠, 99.995% trace metals basis
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Supelco
氯仿, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
甲酸铵 溶液, 10 mM in H2O, suitable for HPLC