Merck
HPLC Analysis of EDTA on SeQuant® ZIC-HILIC
HPLC Analysis of EDTA on SeQuant® ZIC-HILIC application for HPLC
材料
附屬品
产品编号
说明
价格
瓶形喷雾器
capacity 240 mL
矩形 TLC 展开罐,完整
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Sigma 薄层色谱喷雾盒
Spectroline®Spectroline E 系列紫外灯
output8 W, AC/DC input 115 V AC
矩形 TLC 展开罐,完整
L × H × W 7.5 cm × 15.5 cm × 8.0 cm
Hamilton® 薄层色谱注射器
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),
coated tip
Spectroline®Spectroline E 系列紫外灯
output6 W, AC/DC input 120 V AC
Hamilton® 薄层色谱注射器
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Hamilton® 薄层色谱注射器
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),
coated tip
矩形 TLC 展开罐,完整
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
Spectroline®Spectroline E 系列紫外灯
output6 W, AC/DC input 230 V AC
Spectroline®Spectroline E 系列紫外灯
output4 W, AC/DC input 230 V AC
Spectroline®Spectroline E 系列紫外灯
output8 W, AC/DC input 230 V AC
Latch-lid 薄层色谱展开槽
For use with 10 cm × 10 cm plates
Hamilton® 薄层色谱注射器
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Latch-lid 薄层色谱展开槽
For use with 20 cm × 20 cm plates
Spectroline®Spectroline E 系列紫外灯
output4 W, AC/DC input 115 V AC
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
capacity 20 μL, accuracy: ±0.5%
用于薄层层析板的试剂喷雾器
volume 250 mL, Bottle-Type
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 μm, 200 Å (1.50451.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate
0.5 mL/min
pressure
202 psi (1.4 MPa)
detector
210 nm
injection
10 μL
说明
法律資訊
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany