Skip to Content
Merck

Skip To

HNDGAB-220K

MILLIPLEX® Human Neurodegenerative Autoantibody Magnetic Bead Panel

Simultaneously analyze up to 25 autoantibodies in a bead-based assay using Luminex technology, in human serum, plasma, and cerebrospinal fluid (CSF)

Synonym(s):

Luminex® neuro autoantibody assay, Luminex® neurodegenerative autoantibody assay, Millipore neuro autoantibody panel, Millipore neurodegenerative autoantibody panel, neuro autoantibody kit, neuro autoantibody multiplex kit, neurodegenerative autoantibody kit

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116126
NACRES:
NA.84
eCl@ss:
32160701
To order a Milliplex® kit, please search for your analyte of interest.


Product Name

MILLIPLEX® Human Neurodegenerative Autoantibody Magnetic Bead Panel,

Quality Segment

description

Simultaneously analyze up to 25 autoantibodies in a bead-based assay using Luminex® technology, in human serum, plasma, and cerebrospinal fluid (CSF)

product line

Milliplex®

quality

200

species reactivity

human

technique(s)

multiplexing: suitable

input

serum
plasma

detection method

fluorometric (Luminex® xMAP® technology)

shipped in

wet ice

storage temp.

2-8°C

General description

Neurodegenerative autoantibodies are immune system-produced antibodies that mistakenly attack the body′s own nervous system proteins, disrupting synaptic function and causing inflammation and neuronal damage. They target essential proteins like synaptic proteins, neuronal antigens, cell surface receptors, and aggregation-prone proteins. Detecting these autoantibodies in blood or cerebrospinal fluid (CSF) can provide valuable insights into the autoimmune processes linked to neurodegenerative diseases. Research on autoantibodies enhances our understanding of serious neurological disorders and may lead to new therapeutic options.

Application

MILLIPLEX® Qualified assays undergo rigourous assay development, verification, and Quality Control testing to achieve optimal performance. Simultaneously analyze up to 25 neurodegenerative autoantibodies in human serum, plasma, and CSF.

Analytes included: anti-α-synuclein, anti-AChR (acetylcholine receptor), anti-AGNA (Anti-Glial Nuclear Antibody, SOX1), anti-Ago2 (Su), anti-CRYAB (α B-crystallin), anti-AMPA-R, anti-Amphiphysin, anti-ANNA2 (Ri)/NOVA1, anti-AQP4 (aquaporin-4), anti-Aβ42, anti-CRMP5 (Collapsin Response Mediater Protein 5), anti-DPPX, anti-DRD2 (Dopamine D2 Receptor), anti-ELAVL4 (HuD), anti-GFAP (Glial Fibrillary Acidic Protein), anti-MBP (Myelin Basic Protein), anti-C1q, anti-mGluR1, anti-MOG (Myelin Oligodendrocyte Glycoprotein), anti-NF-L (Neurofilament light chain), anti-NMDAR1, anti-NRXN3 (neurexin-3), anti-PCA1 (Yo)/CDR2, anti-PNMA1 (Ma1), and anti-tTau

Assay Characteristics: Refer to kit protocol for assay cross-reactivity and sensitivity.

Features and Benefits

  • Versatile Analyte Selection: Customize your research with the ability to select from a panel of 25 neurodegenerative autoantibodies, allowing for tailored solutions that address specific research questions.
  • All-in-One Kit Format: Each kit includes all necessary reagents and an intuitive protocol, simplifying the assay process and reducing the need for multiple kits, which saves time and resources.
  • High Precision and Specificity: Achieve reliable results with demonstrated precision (<10% intra-assay CV and <15% inter-assay CV) and minimal cross-reactivity, ensuring the accuracy and trustworthiness of your findings.
  • Unique Neurodegenerative Disease Research Tool: This first-of-its-kind assay enables valuable insights into the autoimmune mechanisms underlying neurodegenerative diseases.

Preparation Note

Recommended storage for kit components is 2-8˚C.

Legal Information

Luminex is a registered trademark of Luminex Corporation
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HCYTABG17KHCYTABA17KHCABMAG13K
description

-

description

Configurable Human Cytokine Autoantibody IgG 23-Plex Panel

description

Configurable Human Cytokine Autoantibody IgA 23-Plex Panel

description

Configurable Human Cancer Autoantibody IgG 15-Plex Panel

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

technique(s)

multiplexing: suitable

technique(s)

multiplexing: suitable

technique(s)

multiplexing: suitable

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP® technology)

detection method

fluorometric (Luminex® xMAP® technology)

detection method

fluorometric (Luminex® xMAP® technology)

detection method

fluorometric (Luminex® xMAP® technology)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library