Skip to Content
Merck
Readybag® Half Fraser Broth with Supplements acc. ISO 11290, 62 g, irradiated, For the primary selective enrichment of Listeria sp.

Readybag® Half FRASER broth with supplements

Readybag® Half Fraser Broth with Supplements acc. ISO 11290, 62 g, irradiated, For the primary selective enrichment of Listeria sp.

Products

Product Category
Brand
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique