[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Titration & Karl Fischer Analysis](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/titration-and-karl-fischer)Supelco® SmartChemicals for Titration
# Supelco® SmartChemicals for Titration
__Section Overview__
- [Titration Goes Digital—Connect Your SmartChemicals to Your Titrator](https://www.sigmaaldrich.com#tritration-goes-digital)
- [Seamless Transfer of Titration Data via RFID Tag](https://www.sigmaaldrich.com#seamless-transfer-titration-data)
- [Four Benefits Using SmartChemicals for Your Titration](https://www.sigmaaldrich.com#four-benefits-using-smartchemicals)
- [SmartChemicals Are Compatible With Mettler Toledo Titrators](https://www.sigmaaldrich.com#mettler-toledo-titrators)
- [Demo Video: Titration goes Digital](https://www.sigmaaldrich.com#video)
- [Supelco® SmartChemicals - Product List](https://www.sigmaaldrich.com#related-products)
[Request Information](https://www.sigmaaldrich.com/US/en/campaigns/titration-and-karl-fischer-titration)
## [](https://www.sigmaaldrich.com)Titration Goes Digital—Connect Your SmartChemicals to Your Titrator
A new titration technology, using Supelco® SmartTitrants & Supelco® SmartStandards transfers data seamlessly to the titrator. Just a brief touch on the SmartChemical RFID (Radio Frequency Identification) tag saves time, reduces errors and always ensures maximum data integrity.
Improve your Karl Fischer Titration Process for Water Determination. Learn how to avoid errors, get tips & tricks, and discover seamless data transfer. [Join the on-demand webinar](https://webinar.sepscience.com/form/improve-your-karl-fischer-titration-process-for-water-determination).
## [](https://www.sigmaaldrich.com)Seamless transfer of titration data via RFID tag
Using SmartChemicals eliminates time-consuming steps and errors by transferring data wirelessly and instantly. Titripur® volumetric solutions, Certipur® volumetric standards and all Aquastar® Karl Fischer titrants and standards are embedded with an RFID tag on the label. All relevant data from the Certificate of Analysis are stored on the RFID tag of the SmartChemical
[Download the Technical Poster](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/105/315/smart-chemicals-poster-pittcon.pdf)
### Just a simple touch transfers all data in seconds
All information needed for method execution, result calculation and documentation is seamlessly transferred
- Product name
- Article number
- Lot/batch no
- Concentrations
- Date of release
- Shelf life
When using standards, the following additional information is also transferred:
- Molecular weight
- Supplier name
- Compliance according to pharmacopeias and ISO
- Measurement uncertainty
### Initial opening date – stored on the tag
Previously opened chemicals are also recognized, the initial opening date is stored in the RFID tag and is shown on the touchscreen. Expired chemicals are automatically locked from use.
## [](https://www.sigmaaldrich.com)Four Benefits Using SmartChemicals for Your Titration
- __Secure data transfer__—ensures complete and correct reagent & standard data transfer in the titration software
- __Ease of use__—intuitive operation and convenient data transfer in one touch
- __Extended quality management—__titer determination, shelf life, compliance data, initial opening date, release date
- __Improved efficiency—__fast data transfer - safes time - no manual writing and no four-eyes principle necessary
## [](https://www.sigmaaldrich.com)SmartChemicals are compatible with METTLER TOLEDO Titrators
### Compact Titrator
- G20S
- G10S
- V30S
- V20S
- V10S
### Excellence Titrators
- T9
- T7
- T5
### External SmartChemical Reader
## [](https://www.sigmaaldrich.com)Demo Video: Titration goes Digital
## [](https://www.sigmaaldrich.com)Supelco® SmartChemicals - Product List
\*All products ending with a “3” as last digit, are embedded with an RFID tag
## Titripur® Volumetric Solutions
| | | | |
|--------------------------------------------------------------------|--------------------------------------------------------------------------------------------------|-----------|--------------|
| Product No. | Name | Pack Size | Packaging |
| [1603231003](https://www.sigmaaldrich.com/US/en/product/MM/160323) | Titripur® Acetic acid solution 2.0 mol/L (2.0 N) | 1 L | PE bottle |
| [1022771003](https://www.sigmaaldrich.com/US/en/product/MM/102277) | Titripur® Ammonium cer(IV)nitrate solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1090791003](https://www.sigmaaldrich.com/US/en/product/MM/109079) | Titripur® Ammonium thiocyante solution 0.1 mol/L (0.1N) | 1 L | PE bottle |
| [1603251003](https://www.sigmaaldrich.com/US/en/product/MM/160325) | Titripur® Barium chloride solution 0.05 mol/L (0.1N) | 1 L | PE bottle |
| [1603241003](https://www.sigmaaldrich.com/US/en/product/MM/160324) | Titripur® Barium chloride solution 0.1 mol/L (0.2 N) | 1 L | PE bottle |
| [1603161003](https://www.sigmaaldrich.com/US/en/product/MM/160316) | Titripur® Bromide Bromate solution 0.05 mol/L (0.1N) | 1 L | PE bottle |
| [1090921003](https://www.sigmaaldrich.com/US/en/product/MM/109092) | Titripur® Cer (IV) sulfate solution 0.1 mol/L (0.2 N) | 1 L | PE bottle |
| [1027841003](https://www.sigmaaldrich.com/US/en/product/MM/102784) | Titripur® Cooper sulfate solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1091641003](https://www.sigmaaldrich.com/US/en/product/MM/109164) | Titripur® Hanus solution 0.1 mol/L for determination of iodine number | 1 L | Glass bottle |
| [1154801003](https://www.sigmaaldrich.com/US/en/product/MM/115480) | Titripur® Hyamine1622 solution for determination of anionic tensides 0.004 mol/L | 1 L | PE bottle |
| [1003261003](https://www.sigmaaldrich.com/US/en/product/MM/100326) | Titripur® Hydrochloric acid solution in 2-propanol 0.1 mol/L (0.1 N) | 1 L | Glass bottle |
| [1602381003](https://www.sigmaaldrich.com/US/en/product/MM/160238) | Titripur® Hydrochloric acid solution 0.01 mol/L (0.01 N) | 1 L | PE bottle |
| [1603271003](https://www.sigmaaldrich.com/US/en/product/MM/160327) | Titripur® Hydrochloric acid solution 0.05 mol/L (0.05 N) | 1 L | PE bottle |
| [1090601003](https://www.sigmaaldrich.com/US/en/product/MM/109060) | Titripur® Hydrochloric acid solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1090609013](https://www.sigmaaldrich.com/US/en/product/MM/109060) | Titripur® Hydrochloric acid solution 0.1 mol/L (0.1 N) | 10 L | Titripac |
| [1090604003](https://www.sigmaaldrich.com/US/en/product/MM/109060) | Titripur® Hydrochloric acid solution 0.1 mol/L (0.1 N) | 4 L | Titripac |
| [1131369013](https://www.sigmaaldrich.com/US/en/product/MM/113136) | Titripur® Hydrochloric acid solution 0.3571 mol/L (1/2.8 N) | 10 L | Titripac |
| [1090581003](https://www.sigmaaldrich.com/US/en/product/MM/109058) | Titripur® Hydrochloric acid solution 0.5 mol/L (0.5 N) | 1 L | PE bottle |
| [1090584003](https://www.sigmaaldrich.com/US/en/product/MM/109058) | Titripur® Hydrochloric acid solution 0.5 mol/L (0.5 N) | 4 L | Titripac |
| [1090571003](https://www.sigmaaldrich.com/US/en/product/MM/109057) | Titripur® Hydrochloric acid solution 1.0 mol/L (1.0 N) | 1 L | PE bottle |
| [1090574003](https://www.sigmaaldrich.com/US/en/product/MM/109057) | Titripur® Hydrochloric acid solution 1.0 mol/L (1.0 N) | 4 L | Titripac |
| [1090579013](https://www.sigmaaldrich.com/US/en/product/MM/109057) | Titripur® Hydrochloric acid solution 1.0 mol/L (1.0 N) | 10 L | Titripac |
| [1090631003](https://www.sigmaaldrich.com/US/en/product/MM/109063) | Titripur® Hydrochloric acid solution 2.0 mol/L (2.0 N) | 1 L | PE bottle |
| [1131349013](https://www.sigmaaldrich.com/US/en/product/MM/113134) | Titripur® Hydrochloric acid solution 3.571 mol/L (1/0.28N) | 10 L | Titripac |
| [1603281003](https://www.sigmaaldrich.com/US/en/product/MM/160328) | Titripur® Hydrochloric acid solution 4.0 mol/L (4.0 N) | 1 L | PE bottle |
| [1603371003](https://www.sigmaaldrich.com/US/en/product/MM/160337) | Titripur® Iodid-Iodate solution 1/128 mol/L (1/64 N) | 1 L | PE bottle |
| [1090991003](https://www.sigmaaldrich.com/US/en/product/MM/109099) | Titripur® Iodine solution 0.05 mol/L (0.1 N) (not for North America) | 1 L | Glass bottle |
| [1090991073](https://www.sigmaaldrich.com/US/en/product/MM/109099) | Titripur® Iodine solution 0.05 mol/L (0.1 N) (for North America only) | 1 L | Glass bottle |
| [1602361003](https://www.sigmaaldrich.com/US/en/product/MM/160236) | Titripur® Nitric acid solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1091431003](https://www.sigmaaldrich.com/US/en/product/MM/109143) | Titripur® Mercury(II)nitrate solution 0.05 mol/L (0.1 N) | 1 L | PE bottle |
| [1603291003](https://www.sigmaaldrich.com/US/en/product/MM/160329) | Titripur® Oxalic acid solution 0.05 mol/L (0.1 mol/L) | 1 L | PE bottle |
| [1603301003](https://www.sigmaaldrich.com/US/en/product/MM/160330) | Titripur® Oxalic acid solution 0.005 mol/L (0.01 mol/L) | 1 L | PE bottle |
| [1090651003](https://www.sigmaaldrich.com/US/en/product/MM/109065) | Titripur® Perchloric acid solution in water free acetic acid 0.1 mol/L (0.1 N) | 1 L | Glass bottle |
| [1603081003](https://www.sigmaaldrich.com/US/en/product/MM/160308) | Titripur® Potassium bromate solution 1/60 mol/L (0.1 N) | 1 L | PE bottle |
| [1603361003](https://www.sigmaaldrich.com/US/en/product/MM/160336) | Titripur® Potassium chloride solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1603311003](https://www.sigmaaldrich.com/US/en/product/MM/160331) | Titripur® Potassium chloride solution 1 mol/L (1 N) | 1 L | PE bottle |
| [1603331003](https://www.sigmaaldrich.com/US/en/product/MM/160333) | Titripur® Potassium dichromate solution 1/60 mol/L (0.1 N) | 1 L | Glass bottle |
| [1603331073](https://www.sigmaaldrich.com/US/en/product/MM/160333) | Titripur® Potassium dichromate solution 1/60 mol/L (0.1 N) | 1 L | Glass bottle |
| [1091191003](https://www.sigmaaldrich.com/US/en/product/MM/109119) | Titripur® Potassium dichromate solution 0.02 mol/L | 1 L | Glass bottle |
| [1091181003](https://www.sigmaaldrich.com/US/en/product/MM/109118) | Titripur® Potassium dichromate solution 1/24 mol/L (0.25 N) | 1 L | Glass bottle |
| [1091121003](https://www.sigmaaldrich.com/US/en/product/MM/109112) | Titripur® Potassium hydroxide solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1091151003](https://www.sigmaaldrich.com/US/en/product/MM/109115) | Titripur® Potassium hydroxide solution in ethanol 0.1 mol/L (0.1 N) | 1 L | Glass bottle |
| [1055441003](https://www.sigmaaldrich.com/US/en/product/MM/105544) | Titripur® Potassium hydroxide solution in 2-propanol 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1091141003](https://www.sigmaaldrich.com/US/en/product/MM/109114) | Titripur® Potassium hydroxide solution in ethanol 0.5 mol/L (0.5 N) | 1 L | Glass bottle |
| [1093511003](https://www.sigmaaldrich.com/US/en/product/MM/109351) | Titripur® Potassium hydroxide solution in methanol 0.5 mol/L (0.5 N) | 1 L | Glass bottle |
| [1603341003](https://www.sigmaaldrich.com/US/en/product/MM/160334) | Titripur® Potassium hydroxide solution in methanol 1 mol/L (1 N) | 1 L | Glass bottle |
| [1091081003](https://www.sigmaaldrich.com/US/en/product/MM/109108) | Titripur® Potassium hydroxide solution 1 mol/L (1 N) | 1 L | PE bottle |
| [1091071007](https://www.sigmaaldrich.com/US/en/product/MM/109107) | Titripur® Potassium hydroxide solution max 0.4 ppm Ca 1 mol/L (1 N) | 1 L | PE bottle |
| [1603351003](https://www.sigmaaldrich.com/US/en/product/MM/160335) | Titripur® Potassium iodate solution1/60 mol/L (0.1 N) | 1 L | PE bottle |
| [1603221003](https://www.sigmaaldrich.com/US/en/product/MM/160322) | Titripur® Potassium permanganate solution 0.002 mol/L (0.01 N) | 1 L | Glass bottle |
| [1603211003](https://www.sigmaaldrich.com/US/en/product/MM/160321) | Titripur® Potassium permanganate solution 0.01 mol/L (0.05 N) | 1 L | Glass bottle |
| [1091211003](https://www.sigmaaldrich.com/US/en/product/MM/109121) | Titripur® Potassium permanganate solution standarized with sodium thiosulfate 0.02 mol/L (0.1 N) | 1 L | Glass bottle |
| [1091221003](https://www.sigmaaldrich.com/US/en/product/MM/109122) | Titripur® Potassium permanganate solution standarized with oxalate 0.02 mol/L (0.1 N) | 1 L | Glass bottle |
| [4801602503](https://www.sigmaaldrich.com/US/en/product/MM/480160) | Titripur® Potassium permanganate solution 0.05 mol/L (0.25 N) | 2.5 L | Glass bottle |
| [1117181003](https://www.sigmaaldrich.com/US/en/product/MM/111718) | Titripur® Silver nitrate solution 0.05 mol/L (0.05 N) | 1 L | PE bottle |
| [1090811003](https://www.sigmaaldrich.com/US/en/product/MM/109081) | Titripur® Silver nitrate solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1090814003](https://www.sigmaaldrich.com/US/en/product/MM/109081) | Titripur® Silver nitrate solution 0.1 mol/L (0.1 N) | 4 L | Titripac |
| [1090819013](https://www.sigmaaldrich.com/US/en/product/MM/109081) | Titripur® Silver nitrate solution 0.1 mol/L (0.1 N) | 10 L | Titripac |
| [1090801003](https://www.sigmaaldrich.com/US/en/product/MM/109080) | Titripur® Silver nitrate solution 1 mol/L (1 N) | 1 L | PE bottle |
| [1062771003](https://www.sigmaaldrich.com/US/en/product/MM/106277) | Titripur® Sodum arsenit solution 0.05 mol/L (0.1 N) | 1 L | PE bottle |
| [1603101003](https://www.sigmaaldrich.com/US/en/product/MM/160310) | Titripur® Sodum carbonate solution 0.05 mol/L (0.1 N) | 1 L | PE bottle |
| [1603091003](https://www.sigmaaldrich.com/US/en/product/MM/160309) | Titripur® Sodium hydroxide solution 0.01 mol/L (0.01 N) | 1 L | PE bottle |
| [1603094003](https://www.sigmaaldrich.com/US/en/product/MM/160309) | Titripur® Sodium hydroxide solution 0.01 mol/L (0.01 N) | 4 L | Titripac |
| [1091420503](https://www.sigmaaldrich.com/US/en/product/MM/109142) | Titripur® Sodium hydroxide solution 0.02 mol/L (0.02 N) | 0.5 L | PE bottle |
| [1091411003](https://www.sigmaaldrich.com/US/en/product/MM/109141) | Titripur® Sodium hydroxide solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1091414003](https://www.sigmaaldrich.com/US/en/product/MM/109141) | Titripur® Sodium hydroxide solution 0.1 mol/L (0.1 N) | 4 L | Titripac |
| [1091419013](https://www.sigmaaldrich.com/US/en/product/MM/109141) | Titripur® Sodium hydroxide solution 0.1 mol/L (0.1 N) | 10 L | Titripac |
| [1091401003](https://www.sigmaaldrich.com/US/en/product/MM/109140) | Titripur® Sodium hydroxide solution 0.2 mol/L (0.2 N) | 1 L | PE bottle |
| [1091409013](https://www.sigmaaldrich.com/US/en/product/MM/109140) | Titripur® Sodium hydroxide solution 0.2 mol/L (0.2 N) | 10 L | Titripac |
| [1091391003](https://www.sigmaaldrich.com/US/en/product/MM/109139) | Titripur® Sodium hydroxide solution 0.25 mol/L (0.25 N) | 1 L | PE bottle |
| [1091399013](https://www.sigmaaldrich.com/US/en/product/MM/109139) | Titripur® Sodium hydroxide solution 0.25 mol/L (0.25 N) | 10 L | Titripac |
| [1055951003](https://www.sigmaaldrich.com/US/en/product/MM/105595) | Titripur® Sodium hydroxide solution 0.33 mol/L (0.33 N) | 1 L | PE bottle |
| [1055959013](https://www.sigmaaldrich.com/US/en/product/MM/105595) | Titripur® Sodium hydroxide solution 0.33 mol/L (0.33 N) | 10 L | Titripac |
| [1091381003](https://www.sigmaaldrich.com/US/en/product/MM/109138) | Titripur® Sodium hydroxide solution 0.5 mol/L (0.5 N) | 1 L | PE bottle |
| [1091384003](https://www.sigmaaldrich.com/US/en/product/MM/109138) | Titripur® Sodium hydroxide solution 0.5 mol/L (0.5 N) | 4 L | Titripac |
| [1091389013](https://www.sigmaaldrich.com/US/en/product/MM/109138) | Titripur® Sodium hydroxide solution 0.5 mol/L (0.5 N) | 10 L | Titripac |
| [1091371003](https://www.sigmaaldrich.com/US/en/product/MM/109137) | Titripur® Sodium hydroxide solution 1 mol/L (1 N) | 1 L | PE bottle |
| [1091374003](https://www.sigmaaldrich.com/US/en/product/MM/109137) | Titripur® Sodium hydroxide solution 1 mol/L (1 N) | 4 L | Titripac |
| [1091379013](https://www.sigmaaldrich.com/US/en/product/MM/109137) | Titripur® Sodium hydroxide solution 1 mol/L (1 N) | 10 L | Titripac |
| [1091361003](https://www.sigmaaldrich.com/US/en/product/MM/109136) | Titripur® Sodium hydroxide solution 2 mol/L (2 N) | 1 L | PE bottle |
| [1603181003](https://www.sigmaaldrich.com/US/en/product/MM/160318) | Titripur® Sodium thiosulfate solution 0.01 mol/L (0.01 N) | 1 L | PE bottle |
| [1603111003](https://www.sigmaaldrich.com/US/en/product/MM/160311) | Titripur® Sodium thiosulfate solution 0.05 mol/L (0.05 N) | 1 L | PE bottle |
| [1091471003](https://www.sigmaaldrich.com/US/en/product/MM/109147) | Titripur® Sodium thiosulfate solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
| [1091474003](https://www.sigmaaldrich.com/US/en/product/MM/109147) | Titripur® Sodium thiosulfate solution 0.1 mol/L (0.1 N) | 4 L | Titripac |
| [1091479013](https://www.sigmaaldrich.com/US/en/product/MM/109147) | Titripur® Sodium thiosulfate solution 0.1 mol/L (0.1 N) | 10 L | Titripac |
| [1603121003](https://www.sigmaaldrich.com/US/en/product/MM/160312) | Titripur® Sodium thiosulfate solution 1 mol/L (1 N) | 1 L | PE bottle |
| [1090741003](https://www.sigmaaldrich.com/US/en/product/MM/109074) | Titripur® Sulfuric acid solution 0.05 mol/L (0.1 N) | 1 L | PE bottle |
| [1090749013](https://www.sigmaaldrich.com/US/en/product/MM/109074) | Titripur® Sulfuric acid solution 0.05 mol/L (0.1 N) | 10 L | Titripac |
| [1090731003](https://www.sigmaaldrich.com/US/en/product/MM/109073) | Titripur® Sulfuric acid solution 0.25 mol/L (0.5 N) | 1 L | PE bottle |
| [1090734003](https://www.sigmaaldrich.com/US/en/product/MM/109073) | Titripur® Sulfuric acid solution 0.25 mol/L (0.5 N) | 4 L | Titripac |
| [1090739013](https://www.sigmaaldrich.com/US/en/product/MM/109073) | Titripur® Sulfuric acid solution 0.25 mol/L (0.5 N) | 10 L | Titripac |
| [1090721003](https://www.sigmaaldrich.com/US/en/product/MM/109072) | Titripur® Sulfuric acid solution 0.5 mol/L (1 N) | 1 L | PE bottle |
| [1090724003](https://www.sigmaaldrich.com/US/en/product/MM/109072) | Titripur® Sulfuric acid solution 0.5 mol/L (1 N) | 4 L | Titripac |
| [1090729013](https://www.sigmaaldrich.com/US/en/product/MM/109072) | Titripur® Sulfuric acid solution 0.5 mol/L (1 N) | 10 L | Titripac |
| [1603131003](https://www.sigmaaldrich.com/US/en/product/MM/160313) | Titripur® Sulfuric acid solution 1 mol/L (2 N) | 1 L | PE bottle |
| [4803641003](https://www.sigmaaldrich.com/US/en/product/MM/480364) | Titripur® Sulfuric acid solution 2.5 mol/L (5 N) | 1 L | PE bottle |
| [1091621003](https://www.sigmaaldrich.com/US/en/product/MM/109162) | Titripur® Tetra-n-butyl-ammonium hydroxide solution in 2-propanol/methanol 0.1 mol/L (0.1 N) | 1 L | Glass bottle |
| [1603191003](https://www.sigmaaldrich.com/US/en/product/MM/160319) | Titripur® Titriplex III Solution 0.01 mol/L (not for North America) | 1 L | PE bottle |
| [1603201003](https://www.sigmaaldrich.com/US/en/product/MM/160320) | Titripur® Titriplex III Solution 0.05 mol/L (not for North America) | 1 L | PE bottle |
| [1084311003](https://www.sigmaaldrich.com/US/en/product/MM/108431) | Titripur® Titriplex III Solution 0.1 mol/L (not for North America) | 1 L | PE bottle |
| [1084311073](https://www.sigmaaldrich.com/US/en/product/MM/108431) | Titripur® Titriplex III Solution 0.1 mol/L (for North America only) | 1 L | PE bottle |
| [1084314003](https://www.sigmaaldrich.com/US/en/product/MM/108431) | Titripur® Titriplex III Solution 0.1 mol/L (not for North America) | 4 L | Titripac |
| [1084319013](https://www.sigmaaldrich.com/US/en/product/MM/108431) | Titripur® Titriplex III Solution 0.1 mol/L (not for North America) | 10 L | Titripac |
| [1084471003](https://www.sigmaaldrich.com/US/en/product/MM/108447) | Titripur® Titriplex IV Solution 0.1 mol/L (not for North America) | 1 L | PE bottle |
| [1084474003](https://www.sigmaaldrich.com/US/en/product/MM/108447) | Titripur® Titriplex IV Solution 0.1 mol/L (not for North America) | 4 L | Titripac |
| [1084191003](https://www.sigmaaldrich.com/US/en/product/MM/108419) | Titripur® Titriplex Solution A (not for North America) | 1 L | PE bottle |
| [1084201003](https://www.sigmaaldrich.com/US/en/product/MM/108420) | Titripur® Titriplex solution B 10 mgCaO/L = 1 ml (not for North America) | 1 L | PE bottle |
| [1084209013](https://www.sigmaaldrich.com/US/en/product/MM/108420) | Titripur® Titirplex solution B 10 mgCaO/L = 1 ml (not for North America) | 10 L | Titripac |
| [1084501003](https://www.sigmaaldrich.com/US/en/product/MM/108450) | Titripur® Trifluoromethansulfonic acid in anhydrous acetic acid 0.1 mol/L | 1 L | Glass bottle |
| [1091631003](https://www.sigmaaldrich.com/US/en/product/MM/109163) | Titripur® Wijs solution (iodine monochloride in acetic acid) 0.1 mol/L | 1 L | Glass bottle |
| [1088791003](https://www.sigmaaldrich.com/US/en/product/MM/108879) | Titripur® Zinc sulfate solution 0.1 mol/L (0.1 N) | 1 L | PE bottle |
## Certipur® Volumetric Standards according to ISO 17034 (Certified Reference Materials)
| | | | |
|--------------------------------------------------------------------|---------------------------------------------------|-----------|--------------|
| Product No. | Name | Pack Size | Packaging |
| [1024000083](https://www.sigmaaldrich.com/US/en/product/MM/102400) | Certipur® Potassium hydrogen phthalate CRM | 80 g | Glass bottle |
| [1024010063](https://www.sigmaaldrich.com/US/en/product/MM/102401) | Certipur® Benzoic acid CRM | 60 g | Glass bottle |
| [1024020083](https://www.sigmaaldrich.com/US/en/product/MM/102402) | Certipur® Iron(II) ethylenediammonium sulfate CRM | 80 g | Glass bottle |
| [1024030083](https://www.sigmaaldrich.com/US/en/product/MM/102403) | Certipur® Potassium dichromate CRM | 80 g | Glass bottle |
| [1024040103](https://www.sigmaaldrich.com/US/en/product/MM/102404) | Certipur® Potassium iodate CRM | 100 g | Glass bottle |
| [1024050083](https://www.sigmaaldrich.com/US/en/product/MM/102405) | Certipur® Sodium carbonate CRM | 80 g | Glass bottle |
| [1024060083](https://www.sigmaaldrich.com/US/en/product/MM/102406) | Certipur® Sodium chloride CRM | 80 g | Glass bottle |
| [1024070063](https://www.sigmaaldrich.com/US/en/product/MM/102407) | Certipur® Di-Sodium oxalate CRM | 60 g | Glass bottle |
| [1024080083](https://www.sigmaaldrich.com/US/en/product/MM/102408) | Certipur® TRIS(Hydroxymethyl)aminomethane CRM | 80 g | Glass bottle |
| [1024090103](https://www.sigmaaldrich.com/US/en/product/MM/102409) | Certipur® Zinc CRM | 100 g | Glass bottle |
| [1024100053](https://www.sigmaaldrich.com/US/en/product/MM/102410) | Certipur® Calcium carbonate CRM | 50 g | Glass bottle |
## Aquastar® Karl Fischer Titrants
| | | | |
|--------------------------------------------------------------------|--------------------------------------------------------------------------------------|-----------|--------------|
| Product No. | Name | Pack Size | Packaging |
| [1880011003](https://www.sigmaaldrich.com/US/en/product/MM/188001) | Aquastar® CombiTitrant 1 one component reagents 1 mg H2O/mL (not for North America) | 1 L | Glass bottle |
| [1880011043](https://www.sigmaaldrich.com/US/en/product/MM/188001) | Aquastar® CombiTitrant 1 one component reagents 1 mg H2O/mL (for North America only) | 1 L | Glass bottle |
| [1880021003](https://www.sigmaaldrich.com/US/en/product/MM/188002) | Aquastar® CombiTitrant 2 one component reagents 2 mg H2O/mL (not for North America) | 1 L | Glass bottle |
| [1880021043](https://www.sigmaaldrich.com/US/en/product/MM/188002) | Aquastar® CombiTitrant 2 one component reagents 2 mg H2O/mL (for North America only) | 1 L | Glass bottle |
| [1880051003](https://www.sigmaaldrich.com/US/en/product/MM/188005) | Aquastar® CombiTitrant 5 one component reagents 5 mg H2O/mL | 1 L | Glass bottle |
| [1880052503](https://www.sigmaaldrich.com/US/en/product/MM/188005) | Aquastar® CombiTitrant 5 one component reagents 5 mg H2O/mL | 2.5 L | Glass bottle |
| [1880061003](https://www.sigmaaldrich.com/US/en/product/MM/188006) | Aquastar® CombiTitrant 5 Keto one component reagents 5 mg H2O/mL | 1 L | Glass bottle |
| [1880101003](https://www.sigmaaldrich.com/US/en/product/MM/188010) | Aquastar® Titrant 5 two component reagents 5 mg H2O/mL (not for North America) | 1 L | Glass bottle |
| [1880101043](https://www.sigmaaldrich.com/US/en/product/MM/188010) | Aquastar® Titrant 5 two component reagents 5 mg H2O/mL (for North America only) | 1 L | Glass bottle |
| [1880102503](https://www.sigmaaldrich.com/US/en/product/MM/188010) | Aquastar® Titrant 5 two component reagents 5 mg H2O/mL (not North America) | 2.5 L | Glass bottle |
| [1880102543](https://www.sigmaaldrich.com/US/en/product/MM/188010) | Aquastar® Titrant 5 two component reagents 5 mg H2O/mL (for North America only) | 2.5 L | Glass bottle |
| [1880111003](https://www.sigmaaldrich.com/US/en/product/MM/188011) | Aquastar® Titrant 2 two component reagents 2 mg H2O/mL (not for North America) | 1 L | Glass bottle |
| [1880111043](https://www.sigmaaldrich.com/US/en/product/MM/188011) | Aquastar® Titrant 2 two component reagents 2 mg H2O/mL (for North America only) | 1 L | Glass bottle |
## Aquastar® Water Standards according to ISO 17034 (Certified Reference Materials)
| | | | |
|--------------------------------------------------------------------|-------------------------------------------------------------|-----------|-----------|
| Product No. | Name | Packsize | Packaging |
| [1880510013](https://www.sigmaaldrich.com/US/en/product/MM/188051) | Aquastar® Water standard 0.01 % CRM | 10 x 8 mL | Ampoules |
| [1880510013](https://www.sigmaaldrich.com/US/en/product/MM/188051) | Aquastar® Water standard 0.1 % CRM (not for North America) | 10 x 8 mL | Ampoules |
| [1880510313](https://www.sigmaaldrich.com/US/en/product/MM/188051) | Aquastar® Water standard 0.1 % CRM (for North America only) | 10 x 8 mL | Ampoules |
| [1880520013](https://www.sigmaaldrich.com/US/en/product/MM/188052) | Aquastar® Water standard 1 % CRM (not for North America) | 10 x 8 mL | Ampoules |
| [1880520313](https://www.sigmaaldrich.com/US/en/product/MM/188052) | Aquastar® Water standard 1 % CRM (for North America only)) | 10 x 8 mL | Ampoules |
| [1129390013](https://www.sigmaaldrich.com/US/en/product/MM/112939) | Aquastar® Lactose standard 5 % CRM | 10 g | PE bottle |
| [1066640103](https://www.sigmaaldrich.com/US/en/product/MM/106664) | Aquastar® Sodium tartrate dihydrate | 100 g | PE bottle |
| [1880550013](https://www.sigmaaldrich.com/US/en/product/MM/188055) | Aquastar® Oil standard CRM (15 to 30 ppm water) | 10 x 8 mL | Ampoules |
__Related Articles__
- [Replace Classical Liquid-Liquid Extractions with EXtrelut® NT](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/solid-phase-extraction/extrelut-sle)
- [Importance of Volumetric Solution Standardization](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/calibration-qualification-and-validation/standardizationof-volumetric-solutions)
- [ChemiSphere App for Digital Reagent Data Transfer](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/titration-and-karl-fischer/chemisphere-for-digitized-data-transfer-in-titration)
- [Karl Fischer Titration Tips: Water Content Measurement](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/titration-and-karl-fischer/measuring-water-content-of-samples-that-do-not-easily-release-water)
- [Coulometric Titration - Faster and More Efficient New Aquastar® Coulometric Reagent for Karl Fischer Water Determination](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/titration-and-karl-fischer/protein-binding-determination)
- [Titration 4.0](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/titration-and-karl-fischer/smartchemicals-improve-titration-data)
- [Titrisol® Concentrates](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/titration-and-karl-fischer/titrisol-concentrates)
__Related Product Categories__
- [Titration Reagents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-reagents/titration-reagents)
- [Analytical Reagents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-reagents)
- [Karl Fischer Standards](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/karl-fischer-standards)
- [Karl Fischer Reagents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-reagents/karl-fischer-reagents)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)