[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Organic Reaction Toolbox](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/organic-reaction-toolbox)SynLectro™ Electrolysis Platform
# SynLectro™ Electrolysis Platform
## REACTIONS WITHIN REACH
### With the new SynLectro™ Electrolysis Platform
Electrochemistry for organic synthesis is a technique that has gained popularity due to its ability to design complex small molecules that have previously been impossible to make or find more efficient ways to make known compounds. By using electrical energy to drive chemical change, a targeted section of the molecule can be transformed while leaving the rest of the structure untouched. In addition, the use of electricity replaces toxic and costly chemicals allowing the synthesis to be completed more efficiently, cleaner, and cheaper.
Our new SynLectro™ Electrolysis Platform is designed to simplify and standardize electrochemical organic synthesis at research and batch scale. With our convenient portfolio, there is no need to design or construct your setup anymore. Just choose your kit or pick your components a’la carte and you are ready to explore new electrochemical reactions.
Learn more about each component of the SynLectro™ Electrolysis Platform:
__Figure 1.__Starter Kit 50 Starter Kit
### Starter Kit 50 Starter Kit
Our 50 mL Starter Kit, containing a 50 mL glass cell, a PTFE stopper and sleeve, two electrode holders, and six electrodes (2 graphite, stainless steel, nickel, lead, and zinc), is the perfect way to experiment using electrochemistry for organic synthesis.
### 200 mL Starter Kit
Our 200 mL Starter Kit, containing a 200 mL glass cell, PTFE stopper and sleeve, two electrode holders, and six electrodes (2 graphite, stainless steel, nickel, lead, and zinc), is the perfect way to scale-up reactions using electrochemistry.
Key Features
- No need to self-design equipment for reactions
- Easy-to-use set-up for provides consistency and reproducibility
- Compatible with commercial power supplies
- Supports 2- or 3-electrode setups
- Ensures safety – open system, stopper separates electrical contacts from electrolyte
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Figure 2.__Glass Cells and Stoppers
### Glass Cells and Stoppers
We offer four different cell configurations, a 50 mL cell, a 50 mL jacketed cell, a 200 mL cell, and a 200 mL jacketed cell. Our jacketed cells provide a simple interface to connect an external temperature control system to either heat or cool the electrolyte.
Our PTFE stopper and PTFE sleeve has been designed to separate electrical contacts from electrolyte and work exclusively with our glass cells.
Key Features
- Designed to simplify setup, optimize reactivity, and improve safety
- Side-arm enables connection to reflux condenser or inert gas
- Jacketed option supports thermal control
- Chemical resistant material (PTFE)
- Protects electrical contacts from electrolytes
- Sleeve (optional) prevents seized joints
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Figure 3.__Electrode Holders
### Electrode Holders
We offer two electrode holder options: a stainless-steel electrode holder and a PTFE electrode holder, to connect the power source to the electrode.
Key Features
- Fits snuggly through stopper, holding electrodes firmly in place to create homogenous electric field
- Electrical contact end is compatible with commercial power supplies
- Stainless steel
- PTFE electrode holder prevents damage/scratching to electrode surface - for use with soft electrodes
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Figure 4.__Electrodes
### Electrodes
Choose from 14 different materials:
- Nickel Electrode
- Stainless Steel Electrode
- Copper Electrode
- Tin Electrode
- Lead Electrode
- Leaded Bronze Electrode
- Zinc Electrode
- Magnesium Electrode
- Aluminum Electrode
- Graphite Electrode
- Glassy Carbon Electrode
- Boron-doped Diamond Electrode
- Nickel Foam Electrode
- Graphite Felt Electrode
Key Features
- Each electrode has a dimension of 3x20x60mm, tailored to fit within our cells
- Tailor designed to fit with cells, allowing for optimal active electrode surface
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### SynLectro™ Electrolysis Platform in Action
We’ve compiled representative literature examples using the SynLectro™ platform for electrochemistry:
[Reductive cyclization to access 2H-2-(aryl)-benzo\[d\]-1,2,3-triazoles](https://doi.org/10.1002/chem.201905874)
[Nitrone synthesis from nitroarenes and aldehydes](https://doi.org/10.1039/C8GC00474A)
[Benzyl-aryl cross-coupling](https://doi.org/10.1002/anie.201804997)
[Phenol and anilide couplings](https://doi.org/10.1126/sciadv.aao3920)
__Related Articles__
- [Acid and Base Chart — Table of Acids & Bases](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/acid-base-chart)
- [1-hydrosilatrane](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/1-hydrosilatrane)
- [Aldol Condensation Reaction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/aldol-condensation-reaction)
- [Baeyer-Villiger Oxidation Reaction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/baeyer-villiger-oxidation-reaction)
- [Biginelli Reaction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/biginelli-reaction)
- [Spirocyclic Building Blocks for Scaffold Assembly](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/carreira-spirocycles)
- [Diels–Alder Reaction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/diels-alder-reaction)
__Related Product Categories__
- [Chemical Synthesis](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-synthesis)
- [Catalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts)
- [Synthesis enabling tools](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/synthesis-enabling-tools)
- [Photoreactors](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/synthesis-enabling-tools/photoreactors)
- [Products with sustainability features](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/greener-alternative-products)
- [Electrosynthesis](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/synthesis-enabling-tools/electrosynthesis-platform)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)