Merck
MMVHPPLUG
Stainless Steel Plugfor Milliflex Oasis VHP Pump Outlet
for use with Milliflex Oasis® VHP resistant pump
Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NB.13
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Product Name
Stainless Steel Plugfor Milliflex Oasis VHP Pump Outlet, for use with Milliflex Oasis® VHP resistant pump
packaging
pkg of 1 kit
manufacturer/tradename
Milliflex Oasis®
technique(s)
Rapid Sterility Testing: suitable
application(s)
pharmaceutical
sterility testing
compatibility
for use with Milliflex Oasis® VHP resistant pump
Application
The Milliflex Oasis® VHP resistant pump is designed to perform filtrations of samples inside an isolator. It is suitable for rapid sterility testing in combination with the Milliflex® Rapid System 2.0.
General description
The compact Milliflex Oasis® VHP resistant pump streamlines filtration steps during testing to accelerate and ease the sample preparation. When used with convenient Milliflex Oasis® filtration funnels, the pump and Milliflex Oasis® units comprise an integrated solution for delivering reliable and accurate results. The pump′s small footprint suits tight workspaces and its low profile allow easy access and usage in isolators. The housing and the external parts are VHP resistant.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany